Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 101805991-101806264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLFM3
Entrez Gene ID
118427
Gene Name
olfactomedin 3
Gene Aliases
NOE3, NOELIN3, OPTIMEDIN
UniGene
Hs.484475
Gene Chromosome Location
Chr.1: 101800028-101997234 on Build GRCh38
Transcripts
NM_001288821.1 NM_001288823.1 NM_058170.3 NR_110210.1 NR_110211.1 NR_110212.1 XM_017000240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6057263 c.722G>T p.W241L Chr.1: 101806113-101806113 on Build GRCh38
4554368 c.679G>A p.G227S Chr.1: 101806156-101806156 on Build GRCh38
237880 c.690A>T p.T230T Chr.1: 101806145-101806145 on Build GRCh38
5917309 c.698G>A p.R233K Chr.1: 101806077-101806077 on Build GRCh38
5896563 c.593-3C>T p.? Chr.1: 101806185-101806185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776235811 Chr.1: 101806041-101806041 on Build GRCh38
rs781052092 Chr.1: 101806222-101806222 on Build GRCh38
rs745714574 Chr.1: 101806145-101806145 on Build GRCh38
rs752327892 Chr.1: 101806217-101806217 on Build GRCh38
rs746961447 Chr.1: 101806137-101806137 on Build GRCh38
rs761990679 Chr.1: 101806155-101806155 on Build GRCh38
rs770876302 Chr.1: 101806030-101806030 on Build GRCh38
rs72729629 Chr.1: 101806219-101806219 on Build GRCh38
rs368439922 Chr.1: 101806211-101806211 on Build GRCh38
rs764153070 Chr.1: 101806046-101806046 on Build GRCh38
rs4907955 Chr.1: 101806218-101806218 on Build GRCh38
rs113549195 Chr.1: 101806205-101806205 on Build GRCh38
rs772170561 Chr.1: 101806169-101806169 on Build GRCh38
rs754358302 Chr.1: 101806204-101806204 on Build GRCh38
rs747817734 Chr.1: 101806115-101806115 on Build GRCh38
rs761176709 Chr.1: 101806182-101806182 on Build GRCh38
rs201708966 Chr.1: 101806045-101806045 on Build GRCh38
rs372076237 Chr.1: 101806213-101806213 on Build GRCh38
rs778489824 Chr.1: 101806108-101806108 on Build GRCh38
rs376626442 Chr.1: 101806231-101806231 on Build GRCh38
rs777382296 Chr.1: 101806136-101806136 on Build GRCh38
rs1020427047 Chr.1: 101806238-101806238 on Build GRCh38
rs777281823 Chr.1: 101806235-101806235 on Build GRCh38
rs368510271 Chr.1: 101806057-101806057 on Build GRCh38
rs746916449 Chr.1: 101806026-101806026 on Build GRCh38
rs756564655 Chr.1: 101806058-101806058 on Build GRCh38
rs756225334 Chr.1: 101806171-101806172 on Build GRCh38
rs376156473 Chr.1: 101806103-101806103 on Build GRCh38
rs781042568 Chr.1: 101806144-101806144 on Build GRCh38
rs756826590 Chr.1: 101806221-101806221 on Build GRCh38
rs761937346 Chr.1: 101806051-101806051 on Build GRCh38
rs766744257 Chr.1: 101806067-101806067 on Build GRCh38
rs774627273 Chr.1: 101806047-101806047 on Build GRCh38
rs150207671 Chr.1: 101806146-101806146 on Build GRCh38
rs770931362 Chr.1: 101806141-101806141 on Build GRCh38
rs375617986 Chr.1: 101806237-101806237 on Build GRCh38
rs372272099 Chr.1: 101806202-101806202 on Build GRCh38
rs755368216 Chr.1: 101806107-101806107 on Build GRCh38
rs1000926796 Chr.1: 101806160-101806160 on Build GRCh38
rs773219212 Chr.1: 101806181-101806181 on Build GRCh38
rs144780647 Chr.1: 101806154-101806154 on Build GRCh38
rs1041226972 Chr.1: 101806028-101806028 on Build GRCh38
rs200241392 Chr.1: 101806098-101806098 on Build GRCh38
rs758026066 Chr.1: 101806127-101806127 on Build GRCh38

View Full Product Details