Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 6923412-6923671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D14
Entrez Gene ID
57533
Gene Name
TBC1 domain family member 14
Gene Aliases
-
UniGene
Hs.518611
Gene Chromosome Location
Chr.4: 6909444-7033118 on Build GRCh38
Transcripts
NM_001113361.1 NM_020773.2 XM_005247986.2 XM_006713895.2 XM_011513507.2 XM_011513508.1 XM_017008477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1056845 c.37G>A p.V13I Chr.4: 6923426-6923426 on Build GRCh38
7393106 c.64G>T p.A22S Chr.4: 6923498-6923498 on Build GRCh38
6797172 c.174C>T p.I58I Chr.4: 6923608-6923608 on Build GRCh38
7393105 c.109G>T p.A37S Chr.4: 6923498-6923498 on Build GRCh38
3380972 c.110C>T p.A37V Chr.4: 6923499-6923499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34860182 Chr.4: 6923510-6923510 on Build GRCh38
rs747972149 Chr.4: 6923465-6923465 on Build GRCh38
rs757038790 Chr.4: 6923561-6923561 on Build GRCh38
rs534605304 Chr.4: 6923471-6923471 on Build GRCh38
rs201677970 Chr.4: 6923517-6923517 on Build GRCh38
rs761242173 Chr.4: 6923611-6923611 on Build GRCh38
rs752220496 Chr.4: 6923616-6923616 on Build GRCh38
rs776630489 Chr.4: 6923485-6923485 on Build GRCh38
rs151295415 Chr.4: 6923509-6923509 on Build GRCh38
rs770894126 Chr.4: 6923483-6923483 on Build GRCh38
rs750890802 Chr.4: 6923530-6923530 on Build GRCh38
rs145665500 Chr.4: 6923454-6923454 on Build GRCh38
rs751948984 Chr.4: 6923498-6923498 on Build GRCh38
rs756133689 Chr.4: 6923637-6923637 on Build GRCh38
rs774590557 Chr.4: 6923594-6923594 on Build GRCh38
rs372794265 Chr.4: 6923504-6923504 on Build GRCh38
rs920313271 Chr.4: 6923533-6923533 on Build GRCh38
rs780068290 Chr.4: 6923638-6923638 on Build GRCh38
rs145200625 Chr.4: 6923515-6923515 on Build GRCh38
rs752512167 Chr.4: 6923522-6923522 on Build GRCh38
rs752647966 Chr.4: 6923458-6923458 on Build GRCh38
rs907771920 Chr.4: 6923457-6923457 on Build GRCh38
rs746956643 Chr.4: 6923477-6923477 on Build GRCh38
rs903958844 Chr.4: 6923628-6923628 on Build GRCh38
rs773032848 Chr.4: 6923472-6923472 on Build GRCh38
rs202214482 Chr.4: 6923545-6923545 on Build GRCh38
rs773530418 Chr.4: 6923609-6923609 on Build GRCh38
rs770789355 Chr.4: 6923513-6923513 on Build GRCh38
rs772532768 Chr.4: 6923608-6923608 on Build GRCh38
rs543768521 Chr.4: 6923549-6923549 on Build GRCh38
rs530223501 Chr.4: 6923636-6923636 on Build GRCh38
rs763432658 Chr.4: 6923490-6923490 on Build GRCh38
rs199606822 Chr.4: 6923642-6923642 on Build GRCh38
rs368992523 Chr.4: 6923449-6923449 on Build GRCh38
rs141167484 Chr.4: 6923516-6923516 on Build GRCh38
rs781015384 Chr.4: 6923563-6923563 on Build GRCh38
rs767938002 Chr.4: 6923528-6923528 on Build GRCh38
rs751175855 Chr.4: 6923621-6923621 on Build GRCh38
rs764593549 Chr.4: 6923496-6923496 on Build GRCh38
rs138681879 Chr.4: 6923587-6923587 on Build GRCh38
rs757751419 Chr.4: 6923439-6923439 on Build GRCh38
rs749322594 Chr.4: 6923578-6923578 on Build GRCh38
rs144858395 Chr.4: 6923508-6923508 on Build GRCh38
rs748819925 Chr.4: 6923452-6923452 on Build GRCh38
rs768908454 Chr.4: 6923518-6923518 on Build GRCh38
rs764772194 Chr.4: 6923541-6923541 on Build GRCh38
rs780338469 Chr.4: 6923575-6923575 on Build GRCh38
rs745850476 Chr.4: 6923567-6923567 on Build GRCh38
rs144981670 Chr.4: 6923543-6923543 on Build GRCh38
rs968009659 Chr.4: 6923565-6923565 on Build GRCh38
rs778473001 Chr.4: 6923464-6923464 on Build GRCh38
rs777428014 Chr.4: 6923548-6923548 on Build GRCh38
rs34768850 Chr.4: 6923644-6923644 on Build GRCh38
rs763687892 Chr.4: 6923620-6923620 on Build GRCh38
rs145484437 Chr.4: 6923624-6923624 on Build GRCh38
rs150001164 Chr.4: 6923512-6923512 on Build GRCh38
rs935404049 Chr.4: 6923634-6923634 on Build GRCh38
rs983140818 Chr.4: 6923540-6923540 on Build GRCh38
rs115334884 Chr.4: 6923617-6923617 on Build GRCh38
rs887438873 Chr.4: 6923501-6923501 on Build GRCh38
rs778562700 Chr.4: 6923505-6923505 on Build GRCh38
rs145732986 Chr.4: 6923486-6923486 on Build GRCh38
rs1047692478 Chr.4: 6923447-6923447 on Build GRCh38
rs142845338 Chr.4: 6923635-6923635 on Build GRCh38
rs903861129 Chr.4: 6923503-6923503 on Build GRCh38
rs772183488 Chr.4: 6923649-6923649 on Build GRCh38
rs370410252 Chr.4: 6923602-6923602 on Build GRCh38
rs745943031 Chr.4: 6923560-6923560 on Build GRCh38
rs766861006 Chr.4: 6923612-6923612 on Build GRCh38
rs756989661 Chr.4: 6923537-6923537 on Build GRCh38
rs141012176 Chr.4: 6923499-6923499 on Build GRCh38
rs769849293 Chr.4: 6923572-6923572 on Build GRCh38
rs200312732 Chr.4: 6923599-6923599 on Build GRCh38
rs375266892 Chr.4: 6923641-6923641 on Build GRCh38
rs768012968 Chr.4: 6923500-6923500 on Build GRCh38
rs570309392 Chr.4: 6923645-6923645 on Build GRCh38
rs183069220 Chr.4: 6923527-6923527 on Build GRCh38

View Full Product Details