Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1551986-1552211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRPD
Entrez Gene ID
128646
Gene Name
signal regulatory protein delta
Gene Aliases
PTPNS1L2, dJ576H24.4
UniGene
Hs.729560
Gene Chromosome Location
Chr.20: 1534251-1557697 on Build GRCh38
Transcripts
NM_178460.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5082906 c.117G>A p.Q39Q Chr.20: 1551995-1551995 on Build GRCh38
5973963 c.92A>C p.H31P Chr.20: 1552020-1552020 on Build GRCh38
315202 c.117G>T p.Q39H Chr.20: 1551995-1551995 on Build GRCh38
4781698 c.104C>T p.T35M Chr.20: 1552008-1552008 on Build GRCh38
5676169 c.123A>T p.V41V Chr.20: 1551989-1551989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs118123421 Chr.20: 1552042-1552042 on Build GRCh38
rs760700832 Chr.20: 1552075-1552075 on Build GRCh38
rs777907776 Chr.20: 1552035-1552035 on Build GRCh38
rs16995146 Chr.20: 1552141-1552141 on Build GRCh38
rs749693028 Chr.20: 1552050-1552050 on Build GRCh38
rs758223389 Chr.20: 1552092-1552092 on Build GRCh38
rs765253308 Chr.20: 1552090-1552090 on Build GRCh38
rs1006772253 Chr.20: 1552157-1552157 on Build GRCh38
rs775811912 Chr.20: 1552040-1552040 on Build GRCh38
rs1032937414 Chr.20: 1552171-1552171 on Build GRCh38
rs780841224 Chr.20: 1552022-1552022 on Build GRCh38
rs560159638 Chr.20: 1552165-1552165 on Build GRCh38
rs752383120 Chr.20: 1552028-1552029 on Build GRCh38
rs997206948 Chr.20: 1552121-1552121 on Build GRCh38
rs773257881 Chr.20: 1552071-1552071 on Build GRCh38
rs187691061 Chr.20: 1552098-1552098 on Build GRCh38
rs758372669 Chr.20: 1552158-1552158 on Build GRCh38
rs149087449 Chr.20: 1552140-1552140 on Build GRCh38
rs1026489588 Chr.20: 1552106-1552106 on Build GRCh38
rs866429642 Chr.20: 1552020-1552020 on Build GRCh38
rs770401949 Chr.20: 1552038-1552038 on Build GRCh38
rs145271590 Chr.20: 1552019-1552019 on Build GRCh38
rs752513792 Chr.20: 1552091-1552091 on Build GRCh38
rs112844800 Chr.20: 1552170-1552170 on Build GRCh38
rs17720808 Chr.20: 1552160-1552160 on Build GRCh38
rs768835234 Chr.20: 1552053-1552053 on Build GRCh38
rs957320441 Chr.20: 1552187-1552187 on Build GRCh38
rs756935193 Chr.20: 1552097-1552097 on Build GRCh38
rs751753680 Chr.20: 1552169-1552169 on Build GRCh38
rs914441514 Chr.20: 1552093-1552093 on Build GRCh38
rs540684925 Chr.20: 1552096-1552096 on Build GRCh38
rs1054394932 Chr.20: 1552063-1552063 on Build GRCh38
rs746413734 Chr.20: 1552033-1552033 on Build GRCh38
rs1016524046 Chr.20: 1552159-1552159 on Build GRCh38
rs866634758 Chr.20: 1552041-1552041 on Build GRCh38
rs751475603 Chr.20: 1552070-1552070 on Build GRCh38
rs995458461 Chr.20: 1552103-1552103 on Build GRCh38
rs767625211 Chr.20: 1552066-1552066 on Build GRCh38
rs758217989 Chr.20: 1552043-1552044 on Build GRCh38
rs369940226 Chr.20: 1552065-1552065 on Build GRCh38
rs777582745 Chr.20: 1552053-1552053 on Build GRCh38

View Full Product Details