Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43976263-43976504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0B
Entrez Gene ID
533
Gene Name
ATPase H+ transporting V0 subunit b
Gene Aliases
ATP6F, HATPL, VMA16
UniGene
Hs.596514
Gene Chromosome Location
Chr.1: 43974648-43978300 on Build GRCh38
Transcripts
NM_001039457.2 NM_001294333.1 NM_004047.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4008321 c.235G>A p.G79R Chr.1: 43976336-43976336 on Build GRCh38
5025263 c.276C>T p.V92V Chr.1: 43976377-43976377 on Build GRCh38
6875791 c.225C>T p.S75S Chr.1: 43976326-43976326 on Build GRCh38
214141 c.271C>A p.L91M Chr.1: 43976372-43976372 on Build GRCh38
6394983 c.278G>T p.S93I Chr.1: 43976379-43976379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768805395 Chr.1: 43976409-43976409 on Build GRCh38
rs979009134 Chr.1: 43976461-43976461 on Build GRCh38
rs925137737 Chr.1: 43976467-43976467 on Build GRCh38
rs780705853 Chr.1: 43976311-43976311 on Build GRCh38
rs752866602 Chr.1: 43976282-43976283 on Build GRCh38
rs947478879 Chr.1: 43976387-43976387 on Build GRCh38
rs1052226935 Chr.1: 43976479-43976479 on Build GRCh38
rs772418698 Chr.1: 43976385-43976385 on Build GRCh38
rs757711676 Chr.1: 43976320-43976320 on Build GRCh38
rs926157593 Chr.1: 43976468-43976468 on Build GRCh38
rs761741458 Chr.1: 43976426-43976426 on Build GRCh38
rs1016111717 Chr.1: 43976308-43976308 on Build GRCh38
rs932800773 Chr.1: 43976451-43976451 on Build GRCh38
rs765350813 Chr.1: 43976355-43976355 on Build GRCh38
rs746276423 Chr.1: 43976365-43976365 on Build GRCh38
rs749764900 Chr.1: 43976317-43976317 on Build GRCh38
rs976819968 Chr.1: 43976340-43976340 on Build GRCh38
rs762437462 Chr.1: 43976465-43976465 on Build GRCh38
rs921246807 Chr.1: 43976350-43976350 on Build GRCh38
rs747061836 Chr.1: 43976404-43976404 on Build GRCh38
rs369879383 Chr.1: 43976291-43976291 on Build GRCh38
rs373170197 Chr.1: 43976394-43976394 on Build GRCh38
rs756321064 Chr.1: 43976375-43976383 on Build GRCh38
rs766074669 Chr.1: 43976292-43976292 on Build GRCh38
rs754535852 Chr.1: 43976299-43976299 on Build GRCh38
rs751042342 Chr.1: 43976298-43976298 on Build GRCh38
rs765040488 Chr.1: 43976287-43976287 on Build GRCh38
rs776822634 Chr.1: 43976421-43976421 on Build GRCh38
rs935145608 Chr.1: 43976472-43976472 on Build GRCh38
rs921466399 Chr.1: 43976428-43976428 on Build GRCh38
rs776603858 Chr.1: 43976419-43976419 on Build GRCh38
rs750186065 Chr.1: 43976288-43976288 on Build GRCh38
rs779280296 Chr.1: 43976323-43976323 on Build GRCh38

View Full Product Details