Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119076433-119076660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WARS2
Entrez Gene ID
10352
Gene Name
tryptophanyl tRNA synthetase 2, mitochondrial
Gene Aliases
TrpRS
UniGene
Hs.523506
Gene Chromosome Location
Chr.1: 119031216-119140701 on Build GRCh38
Transcripts
NM_015836.3 NM_201263.2 XM_005270350.3 XM_011540494.2 XM_011540495.2 XM_017000038.1 XM_017000039.1 XM_017000040.1 XM_017000041.1 XM_017000042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5843246 c.233C>T p.S78F Chr.1: 119076465-119076465 on Build GRCh38
7213012 c.155A>G p.Y52C Chr.1: 119076543-119076543 on Build GRCh38
5843248 c.149G>A p.G50D Chr.1: 119076549-119076549 on Build GRCh38
7213013 c.155A>G p.Y52C Chr.1: 119076543-119076543 on Build GRCh38
2179540 c.244C>T p.P82S Chr.1: 119076454-119076454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867578924 Chr.1: 119076579-119076579 on Build GRCh38
rs923069804 Chr.1: 119076493-119076493 on Build GRCh38
rs748018970 Chr.1: 119076589-119076589 on Build GRCh38
rs774688679 Chr.1: 119076468-119076468 on Build GRCh38
rs11552864 Chr.1: 119076550-119076550 on Build GRCh38
rs950496986 Chr.1: 119076497-119076497 on Build GRCh38
rs772271042 Chr.1: 119076633-119076633 on Build GRCh38
rs139975529 Chr.1: 119076551-119076551 on Build GRCh38
rs919278122 Chr.1: 119076609-119076609 on Build GRCh38
rs929530494 Chr.1: 119076623-119076623 on Build GRCh38
rs763093010 Chr.1: 119076463-119076463 on Build GRCh38
rs975735366 Chr.1: 119076481-119076481 on Build GRCh38
rs866710232 Chr.1: 119076495-119076495 on Build GRCh38
rs757986105 Chr.1: 119076578-119076578 on Build GRCh38
rs766501807 Chr.1: 119076467-119076467 on Build GRCh38
rs779551463 Chr.1: 119076580-119076580 on Build GRCh38
rs754687998 Chr.1: 119076582-119076582 on Build GRCh38
rs773290794 Chr.1: 119076455-119076455 on Build GRCh38
rs149515458 Chr.1: 119076499-119076499 on Build GRCh38
rs778193998 Chr.1: 119076592-119076592 on Build GRCh38
rs371009025 Chr.1: 119076469-119076469 on Build GRCh38
rs749569280 Chr.1: 119076619-119076619 on Build GRCh38
rs756704413 Chr.1: 119076519-119076519 on Build GRCh38
rs780986003 Chr.1: 119076587-119076587 on Build GRCh38
rs771086441 Chr.1: 119076626-119076626 on Build GRCh38
rs371681806 Chr.1: 119076581-119076581 on Build GRCh38
rs774400311 Chr.1: 119076629-119076629 on Build GRCh38
rs587742341 Chr.1: 119076591-119076591 on Build GRCh38
rs142739265 Chr.1: 119076454-119076454 on Build GRCh38
rs753210309 Chr.1: 119076510-119076510 on Build GRCh38

View Full Product Details