Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15435587-15435828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF7
Entrez Gene ID
114905
Gene Name
C1q and tumor necrosis factor related protein 7
Gene Aliases
CTRP7, ZACRP7
UniGene
Hs.153714
Gene Chromosome Location
Chr.4: 15339818-15446169 on Build GRCh38
Transcripts
NM_001135170.1 NM_001135171.1 NM_031911.4 XM_011513772.1 XM_011513773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5865899 c.44G>A p.S15N Chr.4: 15435787-15435787 on Build GRCh38
362260 c.55C>A p.R19R Chr.4: 15435798-15435798 on Build GRCh38
4765059 c.56_57insG p.N21fs*1 Chr.4: 15435799-15435800 on Build GRCh38
732355 c.18T>C p.Y6Y Chr.4: 15435761-15435761 on Build GRCh38
1052206 c.21T>C p.V7V Chr.4: 15435764-15435764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755392208 Chr.4: 15435795-15435795 on Build GRCh38
rs565955774 Chr.4: 15435707-15435707 on Build GRCh38
rs754457076 Chr.4: 15435774-15435774 on Build GRCh38
rs755443466 Chr.4: 15435693-15435693 on Build GRCh38
rs372719776 Chr.4: 15435715-15435715 on Build GRCh38
rs770497139 Chr.4: 15435714-15435714 on Build GRCh38
rs764617258 Chr.4: 15435788-15435788 on Build GRCh38
rs777557321 Chr.4: 15435697-15435697 on Build GRCh38
rs890505562 Chr.4: 15435799-15435799 on Build GRCh38
rs766971574 Chr.4: 15435766-15435766 on Build GRCh38
rs1001345092 Chr.4: 15435674-15435674 on Build GRCh38
rs552155873 Chr.4: 15435658-15435658 on Build GRCh38
rs768494606 Chr.4: 15435729-15435729 on Build GRCh38
rs531973801 Chr.4: 15435636-15435636 on Build GRCh38
rs148016047 Chr.4: 15435755-15435755 on Build GRCh38
rs754224424 Chr.4: 15435690-15435690 on Build GRCh38
rs780447221 Chr.4: 15435689-15435689 on Build GRCh38
rs539984171 Chr.4: 15435675-15435675 on Build GRCh38
rs140840563 Chr.4: 15435743-15435743 on Build GRCh38
rs888181336 Chr.4: 15435653-15435653 on Build GRCh38
rs750467017 Chr.4: 15435760-15435760 on Build GRCh38
rs753128594 Chr.4: 15435800-15435800 on Build GRCh38
rs866089493 Chr.4: 15435708-15435708 on Build GRCh38
rs1048143446 Chr.4: 15435618-15435618 on Build GRCh38
rs115546139 Chr.4: 15435617-15435617 on Build GRCh38
rs967385514 Chr.4: 15435746-15435746 on Build GRCh38
rs753448290 Chr.4: 15435625-15435625 on Build GRCh38
rs779357256 Chr.4: 15435798-15435798 on Build GRCh38
rs769735073 Chr.4: 15435718-15435718 on Build GRCh38
rs775442925 Chr.4: 15435723-15435723 on Build GRCh38
rs200945124 Chr.4: 15435727-15435727 on Build GRCh38
rs767571571 Chr.4: 15435747-15435747 on Build GRCh38
rs537297407 Chr.4: 15435762-15435762 on Build GRCh38
rs754468609 Chr.4: 15435734-15435734 on Build GRCh38

View Full Product Details