Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27215348-27215577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRAID
Entrez Gene ID
51374
Gene Name
all-trans retinoic acid induced differentiation factor
Gene Aliases
APR--3, APR-3, APR3, C2orf28, HSPC013, PRO240, p18
UniGene
Hs.9527
Gene Chromosome Location
Chr.2: 27212031-27217178 on Build GRCh38
Transcripts
NM_001170795.1 NM_016085.4 NM_080592.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4093495 c.460G>T p.D154Y Chr.2: 27215475-27215475 on Build GRCh38
3580549 c.477C>T p.P159P Chr.2: 27215492-27215492 on Build GRCh38
5414294 c.529C>T p.L177L Chr.2: 27215544-27215544 on Build GRCh38
7357095 c.487G>A p.D163N Chr.2: 27215502-27215502 on Build GRCh38
5580444 c.500C>T p.T167I Chr.2: 27215515-27215515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117160258 Chr.2: 27215431-27215431 on Build GRCh38
rs760508001 Chr.2: 27215422-27215422 on Build GRCh38
rs748602378 Chr.2: 27215403-27215403 on Build GRCh38
rs147821107 Chr.2: 27215520-27215520 on Build GRCh38
rs199615657 Chr.2: 27215461-27215461 on Build GRCh38
rs760103019 Chr.2: 27215489-27215489 on Build GRCh38
rs778540246 Chr.2: 27215511-27215511 on Build GRCh38
rs766723588 Chr.2: 27215459-27215459 on Build GRCh38
rs144459824 Chr.2: 27215502-27215502 on Build GRCh38
rs144087449 Chr.2: 27215469-27215469 on Build GRCh38
rs533951569 Chr.2: 27215413-27215413 on Build GRCh38
rs753293030 Chr.2: 27215493-27215493 on Build GRCh38
rs781726654 Chr.2: 27215401-27215401 on Build GRCh38
rs770248100 Chr.2: 27215407-27215407 on Build GRCh38
rs763792206 Chr.2: 27215372-27215372 on Build GRCh38
rs775061889 Chr.2: 27215410-27215411 on Build GRCh38
rs780385427 Chr.2: 27215384-27215384 on Build GRCh38
rs778084612 Chr.2: 27215411-27215411 on Build GRCh38
rs999853362 Chr.2: 27215404-27215404 on Build GRCh38
rs756543822 Chr.2: 27215496-27215496 on Build GRCh38
rs781123708 Chr.2: 27215475-27215475 on Build GRCh38
rs754958466 Chr.2: 27215400-27215400 on Build GRCh38
rs867875375 Chr.2: 27215391-27215391 on Build GRCh38
rs776804621 Chr.2: 27215380-27215380 on Build GRCh38
rs984810824 Chr.2: 27215540-27215540 on Build GRCh38
rs201262854 Chr.2: 27215531-27215531 on Build GRCh38
rs771226229 Chr.2: 27215507-27215507 on Build GRCh38
rs761654510 Chr.2: 27215446-27215446 on Build GRCh38
rs774832372 Chr.2: 27215521-27215521 on Build GRCh38
rs747110619 Chr.2: 27215389-27215389 on Build GRCh38
rs147492156 Chr.2: 27215499-27215499 on Build GRCh38
rs763990232 Chr.2: 27215433-27215433 on Build GRCh38
rs747787446 Chr.2: 27215445-27215445 on Build GRCh38
rs752758549 Chr.2: 27215545-27215546 on Build GRCh38
rs767715472 Chr.2: 27215493-27215494 on Build GRCh38
rs200808122 Chr.2: 27215512-27215512 on Build GRCh38
rs368056040 Chr.2: 27215415-27215415 on Build GRCh38
rs775564143 Chr.2: 27215419-27215419 on Build GRCh38
rs771416693 Chr.2: 27215417-27215417 on Build GRCh38
rs560000714 Chr.2: 27215533-27215533 on Build GRCh38

View Full Product Details