Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133239919-133240160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VENTX
Entrez Gene ID
27287
Gene Name
VENT homeobox
Gene Aliases
HPX42B, NA88A, VENTX2
UniGene
Hs.125231
Gene Chromosome Location
Chr.10: 133237904-133241930 on Build GRCh38
Transcripts
NM_014468.3 XM_017016073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3751659 c.571G>A p.G191R Chr.10: 133240100-133240100 on Build GRCh38
7095604 c.631G>C p.E211Q Chr.10: 133240160-133240160 on Build GRCh38
5597872 c.403-1G>A p.? Chr.10: 133239931-133239931 on Build GRCh38
5370386 c.419A>T p.Q140L Chr.10: 133239948-133239948 on Build GRCh38
3436012 c.603C>T p.S201S Chr.10: 133240132-133240132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754269330 Chr.10: 133240113-133240113 on Build GRCh38
rs766645489 Chr.10: 133240112-133240112 on Build GRCh38
rs757886103 Chr.10: 133240028-133240028 on Build GRCh38
rs879126686 Chr.10: 133240017-133240017 on Build GRCh38
rs756849805 Chr.10: 133240108-133240126 on Build GRCh38
rs756488565 Chr.10: 133240011-133240011 on Build GRCh38
rs749679756 Chr.10: 133240132-133240134 on Build GRCh38
rs759933048 Chr.10: 133240122-133240122 on Build GRCh38
rs45620332 Chr.10: 133239954-133239954 on Build GRCh38
rs757903840 Chr.10: 133240124-133240124 on Build GRCh38
rs866974231 Chr.10: 133239957-133239957 on Build GRCh38
rs761901060 Chr.10: 133240008-133240008 on Build GRCh38
rs751261659 Chr.10: 133240041-133240043 on Build GRCh38
rs770778761 Chr.10: 133240067-133240067 on Build GRCh38
rs765876200 Chr.10: 133240123-133240123 on Build GRCh38
rs765090857 Chr.10: 133240002-133240002 on Build GRCh38
rs572087517 Chr.10: 133240084-133240084 on Build GRCh38
rs556426846 Chr.10: 133239981-133239981 on Build GRCh38
rs775363542 Chr.10: 133240087-133240087 on Build GRCh38
rs777333048 Chr.10: 133240127-133240127 on Build GRCh38
rs180762061 Chr.10: 133240062-133240062 on Build GRCh38
rs772872288 Chr.10: 133239985-133239985 on Build GRCh38
rs745603635 Chr.10: 133240083-133240083 on Build GRCh38
rs777332130 Chr.10: 133240035-133240035 on Build GRCh38
rs376205046 Chr.10: 133240132-133240132 on Build GRCh38
rs767616096 Chr.10: 133240099-133240099 on Build GRCh38
rs760949594 Chr.10: 133240103-133240103 on Build GRCh38
rs776607726 Chr.10: 133239996-133239996 on Build GRCh38
rs139394666 Chr.10: 133240009-133240009 on Build GRCh38
rs766841045 Chr.10: 133240018-133240018 on Build GRCh38
rs746659419 Chr.10: 133240044-133240044 on Build GRCh38
rs774425129 Chr.10: 133240004-133240004 on Build GRCh38
rs756915577 Chr.10: 133240135-133240135 on Build GRCh38
rs759463983 Chr.10: 133239997-133239997 on Build GRCh38
rs375001040 Chr.10: 133239953-133239953 on Build GRCh38
rs761834337 Chr.10: 133240093-133240093 on Build GRCh38
rs746811887 Chr.10: 133239987-133239987 on Build GRCh38
rs756457466 Chr.10: 133239948-133239948 on Build GRCh38
rs150884840 Chr.10: 133239956-133239956 on Build GRCh38
rs45485699 Chr.10: 133239971-133239971 on Build GRCh38
rs754465488 Chr.10: 133240021-133240021 on Build GRCh38
rs368423943 Chr.10: 133239972-133239972 on Build GRCh38
rs780771230 Chr.10: 133240140-133240140 on Build GRCh38
rs780634776 Chr.10: 133240126-133240126 on Build GRCh38
rs9418952 Chr.10: 133240100-133240100 on Build GRCh38
rs150065198 Chr.10: 133240023-133240023 on Build GRCh38
rs540901758 Chr.10: 133240024-133240024 on Build GRCh38
rs143679708 Chr.10: 133240070-133240070 on Build GRCh38
rs45619335 Chr.10: 133240015-133240015 on Build GRCh38

View Full Product Details