Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30781762-30782029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF40
Entrez Gene ID
9810
Gene Name
ring finger protein 40
Gene Aliases
BRE1B, RBP95, STARING
UniGene
Hs.65238
Gene Chromosome Location
Chr.16: 30761612-30800231 on Build GRCh38
Transcripts
XM_011545997.1 NM_001080417.1 XM_005255222.3

Gene Symbol
ZNF629
Entrez Gene ID
23361
Gene Name
zinc finger protein 629
Gene Aliases
ZNF, ZNF65
UniGene
Hs.301094
Gene Chromosome Location
Chr.16: 30778449-30787322 on Build GRCh38
Transcripts
XM_011545997.1 NM_001080417.1 XM_005255222.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4792033 c.2299C>G p.P767A Chr.16: 30782029-30782029 on Build GRCh38
6831928 c.2388C>T p.P796P Chr.16: 30781940-30781940 on Build GRCh38
6856333 c.2378A>G p.E793G Chr.16: 30781950-30781950 on Build GRCh38
5513184 c.2382delA p.K794fs*43 Chr.16: 30781946-30781946 on Build GRCh38
3983068 c.2521C>T p.P841S Chr.16: 30781807-30781807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546803261 Chr.16: 30781994-30781994 on Build GRCh38
rs772627166 Chr.16: 30781859-30781859 on Build GRCh38
rs747822488 Chr.16: 30781840-30781840 on Build GRCh38
rs777468111 Chr.16: 30781935-30781935 on Build GRCh38
rs775901850 Chr.16: 30781863-30781863 on Build GRCh38
rs758366365 Chr.16: 30781792-30781792 on Build GRCh38
rs896712512 Chr.16: 30781805-30781805 on Build GRCh38
rs755583166 Chr.16: 30781847-30781847 on Build GRCh38
rs770940887 Chr.16: 30781990-30781990 on Build GRCh38
rs759328129 Chr.16: 30781928-30781928 on Build GRCh38
rs773610715 Chr.16: 30781989-30781989 on Build GRCh38
rs200180312 Chr.16: 30781785-30781785 on Build GRCh38
rs767243293 Chr.16: 30782005-30782005 on Build GRCh38
rs747498426 Chr.16: 30781952-30781952 on Build GRCh38
rs545052675 Chr.16: 30781781-30781781 on Build GRCh38
rs927638924 Chr.16: 30781834-30781834 on Build GRCh38
rs750315508 Chr.16: 30781788-30781788 on Build GRCh38
rs375449943 Chr.16: 30781850-30781850 on Build GRCh38
rs61741652 Chr.16: 30781848-30781848 on Build GRCh38
rs756766073 Chr.16: 30781945-30781945 on Build GRCh38
rs766311889 Chr.16: 30781926-30781926 on Build GRCh38
rs549014661 Chr.16: 30781827-30781827 on Build GRCh38
rs753356441 Chr.16: 30781943-30781943 on Build GRCh38
rs770138564 Chr.16: 30781986-30781986 on Build GRCh38
rs751503342 Chr.16: 30781927-30781927 on Build GRCh38
rs754773366 Chr.16: 30781812-30781812 on Build GRCh38
rs753600241 Chr.16: 30781975-30781975 on Build GRCh38
rs747375449 Chr.16: 30781869-30781869 on Build GRCh38
rs769177179 Chr.16: 30781922-30781923 on Build GRCh38
rs751369784 Chr.16: 30781808-30781808 on Build GRCh38
rs759396232 Chr.16: 30781996-30781996 on Build GRCh38
rs202217592 Chr.16: 30781804-30781804 on Build GRCh38
rs768994259 Chr.16: 30781953-30781953 on Build GRCh38
rs34121355 Chr.16: 30781871-30781872 on Build GRCh38
rs768921277 Chr.16: 30781872-30781872 on Build GRCh38
rs755779055 Chr.16: 30781931-30781931 on Build GRCh38
rs894680143 Chr.16: 30781944-30781944 on Build GRCh38
rs763013970 Chr.16: 30781924-30781924 on Build GRCh38
rs747325317 Chr.16: 30781908-30781909 on Build GRCh38
rs528325235 Chr.16: 30781974-30781974 on Build GRCh38
rs949638310 Chr.16: 30781824-30781824 on Build GRCh38
rs370049845 Chr.16: 30781891-30781891 on Build GRCh38
rs765345943 Chr.16: 30781897-30781897 on Build GRCh38
rs773416331 Chr.16: 30781916-30781916 on Build GRCh38
rs201721937 Chr.16: 30781793-30781793 on Build GRCh38
rs752282000 Chr.16: 30781930-30781930 on Build GRCh38
rs776950510 Chr.16: 30781879-30781879 on Build GRCh38
rs777040666 Chr.16: 30781780-30781780 on Build GRCh38
rs1057312421 Chr.16: 30781865-30781865 on Build GRCh38

View Full Product Details