Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 159034980-159035241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf45
Entrez Gene ID
152940
Gene Name
chromosome 4 open reading frame 45
Gene Aliases
-
UniGene
Hs.415576
Gene Chromosome Location
Chr.4: 158893532-159082883 on Build GRCh38
Transcripts
NM_152543.2 XM_011531671.2 XM_011531672.2 XM_017007808.1 XM_017007809.1 XM_017007810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
296458 c.60_61insT p.T21fs*43 Chr.4: 159035036-159035037 on Build GRCh38
5697938 c.10G>T p.V4L Chr.4: 159035087-159035087 on Build GRCh38
5905219 c.54G>A p.M18I Chr.4: 159035043-159035043 on Build GRCh38
4930251 c.48A>C p.K16N Chr.4: 159035049-159035049 on Build GRCh38
733277 c.49C>G p.Q17E Chr.4: 159035048-159035048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183013241 Chr.4: 159035112-159035112 on Build GRCh38
rs965959977 Chr.4: 159035158-159035158 on Build GRCh38
rs537931857 Chr.4: 159035056-159035056 on Build GRCh38
rs950615847 Chr.4: 159035094-159035094 on Build GRCh38
rs772959297 Chr.4: 159035097-159035097 on Build GRCh38
rs760133489 Chr.4: 159035127-159035127 on Build GRCh38
rs954252257 Chr.4: 159035175-159035175 on Build GRCh38
rs1004801436 Chr.4: 159035095-159035095 on Build GRCh38
rs753048946 Chr.4: 159035020-159035020 on Build GRCh38
rs765979756 Chr.4: 159035113-159035113 on Build GRCh38
rs749437359 Chr.4: 159035041-159035041 on Build GRCh38
rs778098180 Chr.4: 159035037-159035037 on Build GRCh38
rs989608145 Chr.4: 159035192-159035192 on Build GRCh38
rs771094463 Chr.4: 159035043-159035043 on Build GRCh38
rs773668126 Chr.4: 159035125-159035125 on Build GRCh38
rs555546552 Chr.4: 159035171-159035171 on Build GRCh38
rs1027890473 Chr.4: 159035092-159035092 on Build GRCh38
rs147358132 Chr.4: 159035057-159035057 on Build GRCh38
rs539706222 Chr.4: 159035054-159035054 on Build GRCh38
rs761548825 Chr.4: 159035072-159035072 on Build GRCh38
rs776827098 Chr.4: 159035168-159035168 on Build GRCh38
rs768084190 Chr.4: 159035128-159035128 on Build GRCh38
rs756423963 Chr.4: 159035031-159035031 on Build GRCh38
rs535248101 Chr.4: 159035161-159035161 on Build GRCh38
rs76405587 Chr.4: 159035206-159035206 on Build GRCh38
rs769469799 Chr.4: 159035074-159035074 on Build GRCh38
rs978230338 Chr.4: 159035207-159035207 on Build GRCh38
rs902087290 Chr.4: 159035136-159035136 on Build GRCh38
rs781693158 Chr.4: 159035019-159035019 on Build GRCh38
rs753140790 Chr.4: 159035134-159035134 on Build GRCh38
rs756583685 Chr.4: 159035135-159035135 on Build GRCh38
rs768539532 Chr.4: 159035099-159035099 on Build GRCh38
rs915386621 Chr.4: 159035204-159035204 on Build GRCh38
rs375252006 Chr.4: 159035047-159035047 on Build GRCh38
rs746988208 Chr.4: 159035060-159035080 on Build GRCh38
rs750245733 Chr.4: 159035049-159035049 on Build GRCh38
rs569828849 Chr.4: 159035088-159035088 on Build GRCh38

View Full Product Details