Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 560590-560837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNTD2
Entrez Gene ID
256329
Gene Name
lamin tail domain containing 2
Gene Aliases
C11orf35
UniGene
Hs.669395
Gene Chromosome Location
Chr.11: 554850-560780 on Build GRCh38
Transcripts
NM_173573.2 XM_017017478.1 XM_005253149.3 XM_011520389.1 XM_017018360.1

Gene Symbol
RASSF7
Entrez Gene ID
8045
Gene Name
Ras association domain family member 7
Gene Aliases
C11orf13, HRAS1, HRC1
UniGene
Hs.72925
Gene Chromosome Location
Chr.11: 559776-564025 on Build GRCh38
Transcripts
NM_173573.2 XM_017017478.1 XM_005253149.3 XM_011520389.1 XM_017018360.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3687384 c.34G>A p.E12K Chr.11: 560683-560683 on Build GRCh38
4546471 c.3G>C p.M1I Chr.11: 560714-560714 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1026862131 Chr.11: 560671-560671 on Build GRCh38
rs1024079585 Chr.11: 560709-560709 on Build GRCh38
rs1051976748 Chr.11: 560760-560760 on Build GRCh38
rs926966706 Chr.11: 560780-560780 on Build GRCh38
rs867102155 Chr.11: 560810-560810 on Build GRCh38
rs750044556 Chr.11: 560683-560683 on Build GRCh38
rs573422722 Chr.11: 560640-560640 on Build GRCh38
rs936996304 Chr.11: 560801-560801 on Build GRCh38
rs1042285381 Chr.11: 560787-560787 on Build GRCh38
rs544371648 Chr.11: 560723-560723 on Build GRCh38
rs930288770 Chr.11: 560618-560618 on Build GRCh38
rs910235798 Chr.11: 560739-560739 on Build GRCh38
rs995360107 Chr.11: 560669-560669 on Build GRCh38
rs1012808532 Chr.11: 560707-560707 on Build GRCh38
rs71462097 Chr.11: 560802-560803 on Build GRCh38
rs932139517 Chr.11: 560616-560616 on Build GRCh38
rs760135985 Chr.11: 560688-560688 on Build GRCh38
rs903267403 Chr.11: 560679-560680 on Build GRCh38
rs755485607 Chr.11: 560760-560761 on Build GRCh38
rs749327953 Chr.11: 560733-560733 on Build GRCh38
rs530018271 Chr.11: 560762-560762 on Build GRCh38
rs758138143 Chr.11: 560713-560713 on Build GRCh38
rs561079573 Chr.11: 560732-560732 on Build GRCh38
rs866295238 Chr.11: 560769-560769 on Build GRCh38
rs200922487 Chr.11: 560710-560710 on Build GRCh38
rs1035847747 Chr.11: 560685-560685 on Build GRCh38
rs903235805 Chr.11: 560801-560801 on Build GRCh38
rs959842543 Chr.11: 560689-560689 on Build GRCh38
rs767294419 Chr.11: 560674-560674 on Build GRCh38
rs958590363 Chr.11: 560698-560698 on Build GRCh38
rs934106213 Chr.11: 560786-560786 on Build GRCh38
rs545691809 Chr.11: 560648-560648 on Build GRCh38
rs922765035 Chr.11: 560769-560770 on Build GRCh38
rs201586315 Chr.11: 560655-560655 on Build GRCh38
rs781298535 Chr.11: 560714-560714 on Build GRCh38
rs774945108 Chr.11: 560761-560761 on Build GRCh38
rs899185310 Chr.11: 560660-560660 on Build GRCh38
rs940757330 Chr.11: 560766-560766 on Build GRCh38
rs754292151 Chr.11: 560716-560725 on Build GRCh38
rs573337720 Chr.11: 560801-560802 on Build GRCh38
rs922186232 Chr.11: 560748-560748 on Build GRCh38
rs766964739 Chr.11: 560679-560680 on Build GRCh38
rs1049213079 Chr.11: 560634-560634 on Build GRCh38
rs754974561 Chr.11: 560703-560703 on Build GRCh38
rs765955205 Chr.11: 560692-560692 on Build GRCh38
rs769269053 Chr.11: 560645-560645 on Build GRCh38
rs988868256 Chr.11: 560690-560690 on Build GRCh38
rs978616533 Chr.11: 560624-560624 on Build GRCh38
rs752500733 Chr.11: 560712-560712 on Build GRCh38
rs1052247031 Chr.11: 560636-560636 on Build GRCh38
rs769293889 Chr.11: 560734-560734 on Build GRCh38
rs932128402 Chr.11: 560752-560752 on Build GRCh38
rs753266041 Chr.11: 560697-560697 on Build GRCh38
rs998263703 Chr.11: 560676-560676 on Build GRCh38

View Full Product Details