Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 224955024-224955259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH14
Entrez Gene ID
127602
Gene Name
dynein axonemal heavy chain 14
Gene Aliases
C1orf67, Dnahc14, HL-18, HL18
UniGene
Hs.133977
Gene Chromosome Location
Chr.1: 224928422-225399294 on Build GRCh38
Transcripts
NM_001145154.1 NM_001373.1 NM_144989.2 XM_006711735.3 XM_011544057.2 XM_011544058.2 XM_011544059.2 XM_011544061.2 XM_011544062.2 XM_011544063.2 XM_011544064.2 XM_011544065.2 XM_011544066.2 XM_011544067.2 XM_011544069.2 XM_011544070.2 XM_011544071.2 XM_011544072.2 XM_017000295.1 XM_017000296.1 XM_017000297.1 XM_017000298.1 XM_017000300.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4638495 c.195T>C p.S65S Chr.1: 224955076-224955076 on Build GRCh38
4638496 c.726T>C p.S242S Chr.1: 224955076-224955076 on Build GRCh38
3484198 c.685G>A p.E229K Chr.1: 224955035-224955035 on Build GRCh38
904535 c.198G>T p.L66F Chr.1: 224955079-224955079 on Build GRCh38
7364822 c.212C>T p.T71I Chr.1: 224955093-224955093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769288542 Chr.1: 224955111-224955111 on Build GRCh38
rs781122425 Chr.1: 224955128-224955128 on Build GRCh38
rs757348076 Chr.1: 224955119-224955119 on Build GRCh38
rs775140756 Chr.1: 224955058-224955058 on Build GRCh38
rs981512919 Chr.1: 224955180-224955180 on Build GRCh38
rs766953503 Chr.1: 224955082-224955082 on Build GRCh38
rs372589152 Chr.1: 224955079-224955079 on Build GRCh38
rs201863929 Chr.1: 224955200-224955201 on Build GRCh38
rs930311518 Chr.1: 224955220-224955220 on Build GRCh38
rs770536329 Chr.1: 224955094-224955095 on Build GRCh38
rs758681034 Chr.1: 224955114-224955114 on Build GRCh38
rs774051636 Chr.1: 224955078-224955078 on Build GRCh38
rs763683218 Chr.1: 224955075-224955075 on Build GRCh38
rs745794098 Chr.1: 224955145-224955145 on Build GRCh38
rs373178379 Chr.1: 224955117-224955117 on Build GRCh38
rs1044725710 Chr.1: 224955228-224955228 on Build GRCh38
rs755313029 Chr.1: 224955088-224955088 on Build GRCh38
rs769626088 Chr.1: 224955148-224955148 on Build GRCh38
rs548547159 Chr.1: 224955208-224955208 on Build GRCh38
rs762760064 Chr.1: 224955074-224955074 on Build GRCh38
rs920315891 Chr.1: 224955172-224955172 on Build GRCh38
rs369547604 Chr.1: 224955115-224955115 on Build GRCh38
rs189895054 Chr.1: 224955113-224955113 on Build GRCh38
rs375259141 Chr.1: 224955083-224955083 on Build GRCh38
rs757565195 Chr.1: 224955069-224955069 on Build GRCh38

View Full Product Details