Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 137568857-137569112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZIC3
Entrez Gene ID
7547
Gene Name
Zic family member 3
Gene Aliases
HTX, HTX1, VACTERLX, ZNF203
UniGene
Hs.111227
Gene Chromosome Location
Chr.X: 137566187-137577691 on Build GRCh38
Transcripts
NM_003413.3 XM_017029802.1

SNP Detail

SNP ID Chromosome Location
rs763662353 Chr.X: 137568904-137568904 on Build GRCh38
rs748259350 Chr.X: 137569085-137569085 on Build GRCh38
rs780449661 Chr.X: 137569017-137569017 on Build GRCh38
rs387906498 Chr.X: 137569063-137569063 on Build GRCh38
rs104894962 Chr.X: 137569054-137569054 on Build GRCh38
rs756480202 Chr.X: 137569012-137569012 on Build GRCh38
rs201336440 Chr.X: 137568878-137568878 on Build GRCh38
rs964996865 Chr.X: 137569083-137569083 on Build GRCh38
rs745796725 Chr.X: 137569093-137569093 on Build GRCh38
rs769599424 Chr.X: 137568880-137568880 on Build GRCh38
rs755253459 Chr.X: 137569029-137569029 on Build GRCh38
rs1032474510 Chr.X: 137569005-137569005 on Build GRCh38
rs912844888 Chr.X: 137569080-137569080 on Build GRCh38
rs762654669 Chr.X: 137568890-137568890 on Build GRCh38
rs754070677 Chr.X: 137569020-137569020 on Build GRCh38
rs775055002 Chr.X: 137568886-137568886 on Build GRCh38
rs778782258 Chr.X: 137568882-137568883 on Build GRCh38
rs1001368262 Chr.X: 137569002-137569002 on Build GRCh38
rs938761737 Chr.X: 137568990-137568990 on Build GRCh38
rs143990850 Chr.X: 137569035-137569035 on Build GRCh38
rs772237598 Chr.X: 137569086-137569086 on Build GRCh38
rs768036826 Chr.X: 137568987-137568987 on Build GRCh38
rs771995983 Chr.X: 137569089-137569089 on Build GRCh38
rs773879335 Chr.X: 137568999-137568999 on Build GRCh38
rs751075155 Chr.X: 137568921-137568921 on Build GRCh38
rs750905477 Chr.X: 137568996-137568996 on Build GRCh38
rs761258496 Chr.X: 137568950-137568950 on Build GRCh38
rs373535003 Chr.X: 137568877-137568877 on Build GRCh38
rs779128399 Chr.X: 137568876-137568876 on Build GRCh38

View Full Product Details