Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20327807-20328068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA5B1
Entrez Gene ID
127731
Gene Name
von Willebrand factor A domain containing 5B1
Gene Aliases
-
UniGene
Hs.205178
Gene Chromosome Location
Chr.1: 20290907-20360442 on Build GRCh38
Transcripts
NM_001039500.2 XM_006710360.3 XM_011540683.2 XM_011540684.2 XM_011540685.2 XM_011540688.2 XM_011540690.2 XM_011540691.2 XM_011540692.2 XM_011540693.1 XM_011540695.1 XM_011540697.2 XM_011540698.1 XM_017000305.1 XM_017000306.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6894091 c.1248C>A p.Y416* Chr.1: 20327994-20327994 on Build GRCh38
5634095 c.1184G>T p.C395F Chr.1: 20327930-20327930 on Build GRCh38
5135474 c.1195A>G p.I399V Chr.1: 20327941-20327941 on Build GRCh38
5958802 c.1226delT p.P411fs*21 Chr.1: 20327972-20327972 on Build GRCh38
4230052 c.362C>A p.S121Y Chr.1: 20327957-20327957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs267598338 Chr.1: 20327977-20327977 on Build GRCh38
rs1019641828 Chr.1: 20327939-20327939 on Build GRCh38
rs764955204 Chr.1: 20327994-20327994 on Build GRCh38
rs1005113635 Chr.1: 20327897-20327897 on Build GRCh38
rs542177831 Chr.1: 20327920-20327920 on Build GRCh38
rs574886920 Chr.1: 20327911-20327911 on Build GRCh38
rs372295718 Chr.1: 20327916-20327916 on Build GRCh38
rs61739563 Chr.1: 20327970-20327970 on Build GRCh38
rs779951626 Chr.1: 20327917-20327917 on Build GRCh38
rs758304488 Chr.1: 20327891-20327891 on Build GRCh38
rs200749312 Chr.1: 20327838-20327838 on Build GRCh38
rs953765112 Chr.1: 20328037-20328037 on Build GRCh38
rs755194019 Chr.1: 20327949-20327949 on Build GRCh38
rs773216146 Chr.1: 20328031-20328031 on Build GRCh38
rs886457907 Chr.1: 20327885-20327885 on Build GRCh38
rs374203397 Chr.1: 20327876-20327876 on Build GRCh38
rs746822356 Chr.1: 20327947-20327947 on Build GRCh38
rs928544252 Chr.1: 20327987-20327987 on Build GRCh38
rs201876766 Chr.1: 20328017-20328017 on Build GRCh38
rs966172294 Chr.1: 20327969-20327969 on Build GRCh38

View Full Product Details