Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117118134-117118385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM45
Entrez Gene ID
80263
Gene Name
tripartite motif containing 45
Gene Aliases
RNF99
UniGene
Hs.301526
Gene Chromosome Location
Chr.1: 117107711-117121789 on Build GRCh38
Transcripts
NM_001145635.1 NM_025188.3 XM_011542199.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7277427 c.962T>C p.L321P Chr.1: 117118294-117118294 on Build GRCh38
6808270 c.889A>C p.K297Q Chr.1: 117118367-117118367 on Build GRCh38
3710033 c.1075A>T p.K359* Chr.1: 117118181-117118181 on Build GRCh38
2178861 c.1096C>T p.P366S Chr.1: 117118160-117118160 on Build GRCh38
3671360 c.993C>A p.T331T Chr.1: 117118263-117118263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768816132 Chr.1: 117118265-117118265 on Build GRCh38
rs144145161 Chr.1: 117118263-117118263 on Build GRCh38
rs769726857 Chr.1: 117118347-117118347 on Build GRCh38
rs758693171 Chr.1: 117118319-117118320 on Build GRCh38
rs563417817 Chr.1: 117118237-117118237 on Build GRCh38
rs781372425 Chr.1: 117118281-117118281 on Build GRCh38
rs564341449 Chr.1: 117118306-117118306 on Build GRCh38
rs775686503 Chr.1: 117118162-117118162 on Build GRCh38
rs752154079 Chr.1: 117118346-117118346 on Build GRCh38
rs928404537 Chr.1: 117118311-117118311 on Build GRCh38
rs771966724 Chr.1: 117118283-117118283 on Build GRCh38
rs949106171 Chr.1: 117118252-117118252 on Build GRCh38
rs146244405 Chr.1: 117118241-117118241 on Build GRCh38
rs200213016 Chr.1: 117118183-117118183 on Build GRCh38
rs746185391 Chr.1: 117118313-117118314 on Build GRCh38
rs755929464 Chr.1: 117118199-117118199 on Build GRCh38
rs753648574 Chr.1: 117118314-117118314 on Build GRCh38
rs774308173 Chr.1: 117118273-117118273 on Build GRCh38
rs747215598 Chr.1: 117118200-117118200 on Build GRCh38
rs201461171 Chr.1: 117118217-117118217 on Build GRCh38
rs376468762 Chr.1: 117118349-117118349 on Build GRCh38
rs34863850 Chr.1: 117118198-117118198 on Build GRCh38
rs778600878 Chr.1: 117118223-117118223 on Build GRCh38
rs373545989 Chr.1: 117118348-117118348 on Build GRCh38
rs780854456 Chr.1: 117118356-117118357 on Build GRCh38
rs979930208 Chr.1: 117118307-117118307 on Build GRCh38
rs759288759 Chr.1: 117118319-117118319 on Build GRCh38
rs747648745 Chr.1: 117118233-117118233 on Build GRCh38
rs910682043 Chr.1: 117118193-117118193 on Build GRCh38
rs773185734 Chr.1: 117118291-117118291 on Build GRCh38
rs779619891 Chr.1: 117118269-117118271 on Build GRCh38
rs372849454 Chr.1: 117118174-117118174 on Build GRCh38
rs368938343 Chr.1: 117118344-117118344 on Build GRCh38
rs542065929 Chr.1: 117118216-117118216 on Build GRCh38
rs749540671 Chr.1: 117118264-117118264 on Build GRCh38
rs201391609 Chr.1: 117118173-117118173 on Build GRCh38
rs370949428 Chr.1: 117118351-117118351 on Build GRCh38
rs189628686 Chr.1: 117118246-117118246 on Build GRCh38
rs761882470 Chr.1: 117118282-117118282 on Build GRCh38
rs754820049 Chr.1: 117118244-117118244 on Build GRCh38
rs766248177 Chr.1: 117118312-117118312 on Build GRCh38

View Full Product Details