Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99538942-99539213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf17
Entrez Gene ID
84103
Gene Name
chromosome 4 open reading frame 17
Gene Aliases
-
UniGene
Hs.97501
Gene Chromosome Location
Chr.4: 99511004-99542303 on Build GRCh38
Transcripts
NM_032149.2 XM_011532315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2951398 c.639G>A p.W213* Chr.4: 99539173-99539173 on Build GRCh38
5908389 c.632A>G p.K211R Chr.4: 99539166-99539166 on Build GRCh38
4956755 c.674T>A p.I225K Chr.4: 99539208-99539208 on Build GRCh38
732146 c.672G>A p.E224E Chr.4: 99539206-99539206 on Build GRCh38
1198835 c.670G>A p.E224K Chr.4: 99539204-99539204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139282942 Chr.4: 99539003-99539003 on Build GRCh38
rs1040801034 Chr.4: 99539064-99539064 on Build GRCh38
rs1003652238 Chr.4: 99539124-99539124 on Build GRCh38
rs72681975 Chr.4: 99539045-99539045 on Build GRCh38
rs200865620 Chr.4: 99539158-99539158 on Build GRCh38
rs766906065 Chr.4: 99539150-99539150 on Build GRCh38
rs759839030 Chr.4: 99539165-99539165 on Build GRCh38
rs763393564 Chr.4: 99539172-99539172 on Build GRCh38
rs1042401260 Chr.4: 99539079-99539079 on Build GRCh38
rs774986733 Chr.4: 99539137-99539137 on Build GRCh38
rs765511991 Chr.4: 99539167-99539167 on Build GRCh38
rs1012731489 Chr.4: 99539177-99539177 on Build GRCh38
rs772661469 Chr.4: 99539141-99539141 on Build GRCh38
rs1049981751 Chr.4: 99539036-99539036 on Build GRCh38
rs532880731 Chr.4: 99539032-99539032 on Build GRCh38
rs983462731 Chr.4: 99539077-99539077 on Build GRCh38
rs769380244 Chr.4: 99539134-99539134 on Build GRCh38
rs965149125 Chr.4: 99539050-99539050 on Build GRCh38
rs974863018 Chr.4: 99539053-99539053 on Build GRCh38
rs1035500717 Chr.4: 99539154-99539154 on Build GRCh38
rs967006017 Chr.4: 99538972-99538972 on Build GRCh38
rs187987713 Chr.4: 99539138-99539138 on Build GRCh38
rs536894904 Chr.4: 99539048-99539048 on Build GRCh38
rs954719096 Chr.4: 99539065-99539065 on Build GRCh38
rs902508657 Chr.4: 99539097-99539097 on Build GRCh38
rs923247085 Chr.4: 99539057-99539057 on Build GRCh38
rs551253143 Chr.4: 99539038-99539038 on Build GRCh38
rs371495156 Chr.4: 99539143-99539143 on Build GRCh38
rs200900171 Chr.4: 99539168-99539168 on Build GRCh38
rs754195565 Chr.4: 99539160-99539160 on Build GRCh38
rs758456508 Chr.4: 99539175-99539175 on Build GRCh38
rs767950763 Chr.4: 99539192-99539193 on Build GRCh38
rs548487782 Chr.4: 99539125-99539125 on Build GRCh38

View Full Product Details