Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 16987933-16988196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB4
Entrez Gene ID
128954
Gene Name
GRB2 associated binding protein family member 4
Gene Aliases
-
UniGene
Hs.434113
Gene Chromosome Location
Chr.22: 16961936-17008281 on Build GRCh38
Transcripts
NM_001037814.1 XM_011546114.2 XM_011546115.1 XM_011546116.2 XM_011546117.1 XM_017028575.1 XM_017028576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4549958 c.487G>A p.G163R Chr.22: 16988159-16988159 on Build GRCh38
5043541 c.602G>T p.C201F Chr.22: 16988044-16988044 on Build GRCh38
51063 c.630A>C p.A210A Chr.22: 16988016-16988016 on Build GRCh38
5963796 c.487G>T p.G163* Chr.22: 16988159-16988159 on Build GRCh38
1414746 c.607C>T p.P203S Chr.22: 16988039-16988039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761618061 Chr.22: 16988055-16988055 on Build GRCh38
rs774057493 Chr.22: 16988146-16988146 on Build GRCh38
rs545946229 Chr.22: 16988037-16988037 on Build GRCh38
rs560437811 Chr.22: 16988017-16988017 on Build GRCh38
rs781008080 Chr.22: 16988108-16988108 on Build GRCh38
rs769659484 Chr.22: 16988020-16988020 on Build GRCh38
rs948805529 Chr.22: 16987965-16987965 on Build GRCh38
rs556202825 Chr.22: 16988005-16988005 on Build GRCh38
rs775507717 Chr.22: 16988127-16988127 on Build GRCh38
rs1045885030 Chr.22: 16987979-16987979 on Build GRCh38
rs878958881 Chr.22: 16988063-16988063 on Build GRCh38
rs766747027 Chr.22: 16988068-16988068 on Build GRCh38
rs61743894 Chr.22: 16987996-16987996 on Build GRCh38
rs777152427 Chr.22: 16988130-16988130 on Build GRCh38
rs201557251 Chr.22: 16988058-16988058 on Build GRCh38
rs772940557 Chr.22: 16988173-16988173 on Build GRCh38
rs770753228 Chr.22: 16988010-16988010 on Build GRCh38
rs376657256 Chr.22: 16987962-16987962 on Build GRCh38
rs779032941 Chr.22: 16988093-16988093 on Build GRCh38
rs749508245 Chr.22: 16987955-16987955 on Build GRCh38
rs762500758 Chr.22: 16988033-16988033 on Build GRCh38
rs763795980 Chr.22: 16987991-16987991 on Build GRCh38
rs200048737 Chr.22: 16988038-16988038 on Build GRCh38
rs751276793 Chr.22: 16987994-16987994 on Build GRCh38
rs759457417 Chr.22: 16987967-16987967 on Build GRCh38
rs1032243450 Chr.22: 16988084-16988084 on Build GRCh38
rs776565964 Chr.22: 16987963-16987963 on Build GRCh38
rs754877111 Chr.22: 16987999-16987999 on Build GRCh38
rs201740103 Chr.22: 16988011-16988011 on Build GRCh38
rs200944025 Chr.22: 16988166-16988166 on Build GRCh38
rs5992604 Chr.22: 16988136-16988136 on Build GRCh38
rs749239188 Chr.22: 16988135-16988135 on Build GRCh38
rs574636837 Chr.22: 16988007-16988007 on Build GRCh38
rs756537092 Chr.22: 16988059-16988059 on Build GRCh38
rs767042060 Chr.22: 16988169-16988169 on Build GRCh38
rs200236379 Chr.22: 16987985-16987985 on Build GRCh38
rs762662155 Chr.22: 16987986-16987986 on Build GRCh38
rs199813045 Chr.22: 16988003-16988003 on Build GRCh38
rs372247155 Chr.22: 16988076-16988076 on Build GRCh38
rs760414370 Chr.22: 16987960-16987960 on Build GRCh38
rs754984849 Chr.22: 16988077-16988077 on Build GRCh38
rs758596697 Chr.22: 16988104-16988104 on Build GRCh38
rs780155519 Chr.22: 16988000-16988000 on Build GRCh38
rs757635484 Chr.22: 16987995-16987995 on Build GRCh38
rs144427521 Chr.22: 16988028-16988028 on Build GRCh38
rs745610221 Chr.22: 16988114-16988114 on Build GRCh38
rs769648537 Chr.22: 16988126-16988126 on Build GRCh38
rs146153837 Chr.22: 16987983-16987983 on Build GRCh38
rs748354526 Chr.22: 16988098-16988098 on Build GRCh38
rs373947928 Chr.22: 16988056-16988056 on Build GRCh38
rs770727743 Chr.22: 16987961-16987961 on Build GRCh38
rs200250480 Chr.22: 16987959-16987959 on Build GRCh38
rs531427759 Chr.22: 16987958-16987958 on Build GRCh38
rs569147492 Chr.22: 16988030-16988030 on Build GRCh38
rs28502153 Chr.22: 16988159-16988159 on Build GRCh38

View Full Product Details