Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54276780-54277009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LILRB2
Entrez Gene ID
10288
Gene Name
leukocyte immunoglobulin like receptor B2
Gene Aliases
CD85D, ILT-4, ILT4, LIR-2, LIR2, MIR-10, MIR10
UniGene
Hs.655652
Gene Chromosome Location
Chr.19: 54273821-54281178 on Build GRCh38
Transcripts
NM_001080978.3 NM_001278403.2 NM_001278404.2 NM_001278405.2 NM_005874.4 NR_103521.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7375055 c.1423C>A p.L475I Chr.19: 54276864-54276864 on Build GRCh38
6701151 c.1456C>T p.R486C Chr.19: 54276831-54276831 on Build GRCh38
245443 c.1458T>C p.R486R Chr.19: 54276829-54276829 on Build GRCh38
3960463 c.1419C>T p.L473L Chr.19: 54276868-54276868 on Build GRCh38
6151996 c.1374G>T p.L458L Chr.19: 54276913-54276913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555858536 Chr.19: 54276978-54276978 on Build GRCh38
rs373805149 Chr.19: 54276888-54276888 on Build GRCh38
rs200943350 Chr.19: 54276885-54276885 on Build GRCh38
rs755006929 Chr.19: 54276958-54276958 on Build GRCh38
rs768865193 Chr.19: 54276831-54276831 on Build GRCh38
rs111762694 Chr.19: 54276833-54276833 on Build GRCh38
rs780260904 Chr.19: 54276853-54276855 on Build GRCh38
rs780161616 Chr.19: 54276813-54276813 on Build GRCh38
rs529493575 Chr.19: 54276798-54276798 on Build GRCh38
rs746360008 Chr.19: 54276840-54276840 on Build GRCh38
rs749515031 Chr.19: 54276830-54276830 on Build GRCh38
rs748450466 Chr.19: 54276843-54276843 on Build GRCh38
rs368141237 Chr.19: 54276879-54276879 on Build GRCh38
rs748538639 Chr.19: 54276901-54276901 on Build GRCh38
rs755209676 Chr.19: 54276900-54276900 on Build GRCh38
rs202071158 Chr.19: 54276954-54276954 on Build GRCh38
rs189835390 Chr.19: 54276951-54276951 on Build GRCh38
rs201245680 Chr.19: 54276977-54276977 on Build GRCh38
rs547037457 Chr.19: 54276909-54276909 on Build GRCh38
rs201896387 Chr.19: 54276882-54276882 on Build GRCh38
rs370944249 Chr.19: 54276806-54276806 on Build GRCh38
rs890903359 Chr.19: 54276853-54276861 on Build GRCh38
rs958854271 Chr.19: 54276959-54276959 on Build GRCh38
rs767258640 Chr.19: 54276940-54276940 on Build GRCh38
rs768455955 Chr.19: 54276937-54276937 on Build GRCh38
rs139961541 Chr.19: 54276915-54276915 on Build GRCh38
rs753943412 Chr.19: 54276897-54276897 on Build GRCh38
rs759104994 Chr.19: 54276854-54276854 on Build GRCh38
rs370398064 Chr.19: 54276886-54276886 on Build GRCh38
rs772568213 Chr.19: 54276844-54276844 on Build GRCh38
rs770393240 Chr.19: 54276852-54276853 on Build GRCh38
rs934955653 Chr.19: 54276852-54276852 on Build GRCh38
rs778175152 Chr.19: 54276907-54276907 on Build GRCh38
rs149508154 Chr.19: 54276808-54276808 on Build GRCh38
rs374388986 Chr.19: 54276939-54276939 on Build GRCh38
rs756050207 Chr.19: 54276887-54276887 on Build GRCh38
rs771464158 Chr.19: 54276913-54276913 on Build GRCh38
rs758711654 Chr.19: 54276906-54276906 on Build GRCh38
rs774293321 Chr.19: 54276883-54276883 on Build GRCh38
rs769235833 Chr.19: 54276855-54276855 on Build GRCh38
rs374263221 Chr.19: 54276809-54276809 on Build GRCh38
rs749421385 Chr.19: 54276853-54276858 on Build GRCh38
rs775032180 Chr.19: 54276864-54276864 on Build GRCh38
rs774098605 Chr.19: 54276938-54276938 on Build GRCh38
rs773059139 Chr.19: 54276944-54276944 on Build GRCh38
rs760591793 Chr.19: 54276945-54276945 on Build GRCh38
rs748749721 Chr.19: 54276928-54276928 on Build GRCh38
rs141786258 Chr.19: 54276881-54276881 on Build GRCh38
rs773549782 Chr.19: 54276849-54276849 on Build GRCh38

View Full Product Details