Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7994887-7995160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3F
Entrez Gene ID
8665
Gene Name
eukaryotic translation initiation factor 3 subunit F
Gene Aliases
EIF3S5, eIF3-p47
UniGene
Hs.516023
Gene Chromosome Location
Chr.11: 7987320-7996171 on Build GRCh38
Transcripts
NM_003754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420224 c.751C>A p.L251M Chr.11: 7994987-7994987 on Build GRCh38
3953852 c.819C>A p.S273R Chr.11: 7995010-7995010 on Build GRCh38
5164735 c.878G>A p.R293H Chr.11: 7995069-7995069 on Build GRCh38
5142184 c.819C>T p.S273S Chr.11: 7995010-7995010 on Build GRCh38
5718960 c.789T>A p.I263I Chr.11: 7995025-7995025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007208285 Chr.11: 7995077-7995077 on Build GRCh38
rs776597083 Chr.11: 7995112-7995112 on Build GRCh38
rs962745284 Chr.11: 7994930-7994930 on Build GRCh38
rs748528533 Chr.11: 7995135-7995139 on Build GRCh38
rs753092228 Chr.11: 7995059-7995059 on Build GRCh38
rs780497184 Chr.11: 7994936-7994937 on Build GRCh38
rs745504983 Chr.11: 7994936-7994936 on Build GRCh38
rs772971739 Chr.11: 7995109-7995109 on Build GRCh38
rs1009667887 Chr.11: 7995078-7995078 on Build GRCh38
rs769093059 Chr.11: 7994977-7994978 on Build GRCh38
rs376830068 Chr.11: 7995087-7995087 on Build GRCh38
rs757615179 Chr.11: 7994915-7994915 on Build GRCh38
rs759496049 Chr.11: 7995044-7995044 on Build GRCh38
rs904105385 Chr.11: 7995069-7995069 on Build GRCh38
rs779325012 Chr.11: 7995133-7995133 on Build GRCh38
rs758099526 Chr.11: 7995082-7995082 on Build GRCh38
rs1056921704 Chr.11: 7995005-7995005 on Build GRCh38
rs374271988 Chr.11: 7995052-7995052 on Build GRCh38
rs759426288 Chr.11: 7995116-7995116 on Build GRCh38
rs771914665 Chr.11: 7994941-7994941 on Build GRCh38
rs199991205 Chr.11: 7995035-7995035 on Build GRCh38
rs748019865 Chr.11: 7995100-7995100 on Build GRCh38
rs778298586 Chr.11: 7994932-7994932 on Build GRCh38
rs377708679 Chr.11: 7995061-7995061 on Build GRCh38
rs567330284 Chr.11: 7995015-7995015 on Build GRCh38
rs56392532 Chr.11: 7994952-7994952 on Build GRCh38
rs1032496529 Chr.11: 7995030-7995030 on Build GRCh38
rs779677506 Chr.11: 7994945-7994945 on Build GRCh38
rs370947755 Chr.11: 7994978-7994978 on Build GRCh38
rs868162509 Chr.11: 7995066-7995066 on Build GRCh38
rs761813296 Chr.11: 7994973-7994973 on Build GRCh38
rs780766725 Chr.11: 7995098-7995098 on Build GRCh38
rs768231585 Chr.11: 7994960-7994960 on Build GRCh38
rs776392059 Chr.11: 7994972-7994972 on Build GRCh38
rs756745169 Chr.11: 7995068-7995068 on Build GRCh38
rs1000975751 Chr.11: 7994961-7994961 on Build GRCh38
rs746533870 Chr.11: 7995099-7995099 on Build GRCh38
rs754291747 Chr.11: 7995076-7995076 on Build GRCh38
rs374099131 Chr.11: 7995101-7995101 on Build GRCh38
rs950577723 Chr.11: 7994917-7994917 on Build GRCh38
rs200650004 Chr.11: 7994995-7994995 on Build GRCh38
rs774373608 Chr.11: 7995133-7995133 on Build GRCh38
rs369290417 Chr.11: 7995090-7995090 on Build GRCh38
rs749770687 Chr.11: 7994942-7994944 on Build GRCh38
rs1033586880 Chr.11: 7995132-7995132 on Build GRCh38
rs984734712 Chr.11: 7994923-7994923 on Build GRCh38
rs773047384 Chr.11: 7994980-7994980 on Build GRCh38
rs754378933 Chr.11: 7995096-7995096 on Build GRCh38

View Full Product Details