Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 111123267-111123492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMMP2L
Entrez Gene ID
83943
Gene Name
inner mitochondrial membrane peptidase subunit 2
Gene Aliases
IMMP2L-IT1, IMP2, IMP2-LIKE
UniGene
Hs.655722
Gene Chromosome Location
Chr.7: 110662645-111562531 on Build GRCh38
Transcripts
NM_001244606.1 NM_032549.3 XM_005250630.3 XM_011516605.2 XM_011516608.2 XM_011516609.2 XM_011516613.2 XM_017012699.1 XM_017012700.1 XM_017012701.1 XM_017012702.1 XM_017012703.1 XM_017012704.1 XM_017012705.1 NM_001099658.1 NM_001099660.1 NM_018334.4

Gene Symbol
LRRN3
Entrez Gene ID
54674
Gene Name
leucine rich repeat neuronal 3
Gene Aliases
FIGLER5, NLRR-3, NLRR3
UniGene
Hs.3781
Gene Chromosome Location
Chr.7: 111091006-111125454 on Build GRCh38
Transcripts
NM_001244606.1 NM_032549.3 XM_005250630.3 XM_011516605.2 XM_011516608.2 XM_011516609.2 XM_011516613.2 XM_017012699.1 XM_017012700.1 XM_017012701.1 XM_017012702.1 XM_017012703.1 XM_017012704.1 XM_017012705.1 NM_001099658.1 NM_001099660.1 NM_018334.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5440713 c.658G>A p.A220T Chr.7: 111123430-111123430 on Build GRCh38
201078 c.690C>T p.N230N Chr.7: 111123462-111123462 on Build GRCh38
230126 c.502C>T p.R168* Chr.7: 111123274-111123274 on Build GRCh38
5986367 c.613G>C p.D205H Chr.7: 111123385-111123385 on Build GRCh38
1256927 c.518C>A p.S173* Chr.7: 111123290-111123290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004357844 Chr.7: 111123424-111123424 on Build GRCh38
rs781214401 Chr.7: 111123369-111123369 on Build GRCh38
rs770573988 Chr.7: 111123440-111123440 on Build GRCh38
rs1039189238 Chr.7: 111123309-111123309 on Build GRCh38
rs35229264 Chr.7: 111123447-111123447 on Build GRCh38
rs769370648 Chr.7: 111123451-111123451 on Build GRCh38
rs760301401 Chr.7: 111123386-111123386 on Build GRCh38
rs536637511 Chr.7: 111123345-111123345 on Build GRCh38
rs773818533 Chr.7: 111123325-111123325 on Build GRCh38
rs563286461 Chr.7: 111123407-111123407 on Build GRCh38
rs1029878494 Chr.7: 111123377-111123377 on Build GRCh38
rs769240546 Chr.7: 111123409-111123409 on Build GRCh38
rs951604736 Chr.7: 111123415-111123415 on Build GRCh38
rs1035980047 Chr.7: 111123356-111123356 on Build GRCh38
rs146578143 Chr.7: 111123353-111123353 on Build GRCh38
rs762758462 Chr.7: 111123453-111123453 on Build GRCh38
rs756359844 Chr.7: 111123374-111123374 on Build GRCh38
rs267601235 Chr.7: 111123423-111123423 on Build GRCh38
rs200291058 Chr.7: 111123416-111123416 on Build GRCh38
rs761798162 Chr.7: 111123463-111123463 on Build GRCh38
rs901551236 Chr.7: 111123348-111123348 on Build GRCh38
rs759192602 Chr.7: 111123311-111123311 on Build GRCh38
rs146797927 Chr.7: 111123462-111123462 on Build GRCh38
rs757334230 Chr.7: 111123359-111123359 on Build GRCh38
rs750645851 Chr.7: 111123372-111123372 on Build GRCh38
rs749666399 Chr.7: 111123400-111123400 on Build GRCh38
rs780437672 Chr.7: 111123387-111123387 on Build GRCh38
rs765065226 Chr.7: 111123318-111123318 on Build GRCh38
rs921598665 Chr.7: 111123458-111123458 on Build GRCh38
rs762844497 Chr.7: 111123337-111123337 on Build GRCh38
rs997554194 Chr.7: 111123362-111123362 on Build GRCh38
rs145004274 Chr.7: 111123454-111123454 on Build GRCh38

View Full Product Details