Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141489135-141489390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA10
Entrez Gene ID
56106
Gene Name
protocadherin gamma subfamily A, 10
Gene Aliases
PCDH-GAMMA-A10
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141413176-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA11
Entrez Gene ID
56105
Gene Name
protocadherin gamma subfamily A, 11
Gene Aliases
PCDH-GAMMA-A11
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141420970-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA12
Entrez Gene ID
26025
Gene Name
protocadherin gamma subfamily A, 12
Gene Aliases
CDH21, FIB3, PCDH-GAMMA-A12
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141430591-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA9
Entrez Gene ID
56107
Gene Name
protocadherin gamma subfamily A, 9
Gene Aliases
PCDH-GAMMA-A9
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141402953-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141398128-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB6
Entrez Gene ID
56100
Gene Name
protocadherin gamma subfamily B, 6
Gene Aliases
PCDH-GAMMA-B6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141408203-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB7
Entrez Gene ID
56099
Gene Name
protocadherin gamma subfamily B, 7
Gene Aliases
ME6, PCDH-GAMMA-B7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141417647-141512981 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGC3
Entrez Gene ID
5098
Gene Name
protocadherin gamma subfamily C, 3
Gene Aliases
PC43, PCDH-GAMMA-C3, PCDH2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141476002-141512977 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGC4
Entrez Gene ID
56098
Gene Name
protocadherin gamma subfamily C, 4
Gene Aliases
PCDH-GAMMA-C4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141485174-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGC5
Entrez Gene ID
56097
Gene Name
protocadherin gamma subfamily C, 5
Gene Aliases
PCDH-GAMMA-C5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141489241-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6587943 c.138G>A p.G46G Chr.5: 141489378-141489378 on Build GRCh38
6253775 c.105G>T p.V35V Chr.5: 141489345-141489345 on Build GRCh38
1063408 c.95G>A p.R32H Chr.5: 141489335-141489335 on Build GRCh38
3208676 c.123G>A p.P41P Chr.5: 141489363-141489363 on Build GRCh38
7313432 c.68G>T p.C23F Chr.5: 141489308-141489308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377248872 Chr.5: 141489231-141489231 on Build GRCh38
rs889945954 Chr.5: 141489368-141489368 on Build GRCh38
rs145484133 Chr.5: 141489244-141489244 on Build GRCh38
rs374235290 Chr.5: 141489314-141489314 on Build GRCh38
rs773391205 Chr.5: 141489245-141489245 on Build GRCh38
rs775028354 Chr.5: 141489230-141489230 on Build GRCh38
rs370726160 Chr.5: 141489267-141489267 on Build GRCh38
rs767837736 Chr.5: 141489363-141489363 on Build GRCh38
rs756528191 Chr.5: 141489294-141489294 on Build GRCh38
rs748457785 Chr.5: 141489197-141489197 on Build GRCh38
rs768509409 Chr.5: 141489236-141489236 on Build GRCh38
rs769607720 Chr.5: 141489346-141489346 on Build GRCh38
rs918375556 Chr.5: 141489318-141489318 on Build GRCh38
rs768635851 Chr.5: 141489334-141489334 on Build GRCh38
rs141115698 Chr.5: 141489246-141489246 on Build GRCh38
rs573088236 Chr.5: 141489279-141489279 on Build GRCh38
rs576983336 Chr.5: 141489165-141489165 on Build GRCh38
rs763097687 Chr.5: 141489361-141489361 on Build GRCh38
rs906371381 Chr.5: 141489181-141489191 on Build GRCh38
rs942218118 Chr.5: 141489180-141489180 on Build GRCh38
rs771766565 Chr.5: 141489214-141489215 on Build GRCh38
rs368330318 Chr.5: 141489329-141489329 on Build GRCh38
rs146919978 Chr.5: 141489268-141489268 on Build GRCh38
rs773010251 Chr.5: 141489354-141489354 on Build GRCh38
rs774363104 Chr.5: 141489365-141489365 on Build GRCh38
rs768135284 Chr.5: 141489277-141489277 on Build GRCh38
rs184934961 Chr.5: 141489297-141489297 on Build GRCh38
rs766390887 Chr.5: 141489250-141489250 on Build GRCh38
rs758024896 Chr.5: 141489313-141489313 on Build GRCh38
rs200660227 Chr.5: 141489195-141489195 on Build GRCh38
rs138015049 Chr.5: 141489335-141489335 on Build GRCh38
rs745541067 Chr.5: 141489312-141489312 on Build GRCh38
rs909255357 Chr.5: 141489178-141489178 on Build GRCh38
rs771455540 Chr.5: 141489223-141489223 on Build GRCh38
rs773157586 Chr.5: 141489257-141489258 on Build GRCh38
rs757039294 Chr.5: 141489201-141489201 on Build GRCh38
rs909780010 Chr.5: 141489255-141489255 on Build GRCh38
rs371328808 Chr.5: 141489369-141489369 on Build GRCh38
rs776473329 Chr.5: 141489241-141489241 on Build GRCh38
rs746520513 Chr.5: 141489323-141489323 on Build GRCh38
rs372898969 Chr.5: 141489213-141489213 on Build GRCh38
rs377697321 Chr.5: 141489343-141489343 on Build GRCh38
rs868211560 Chr.5: 141489289-141489289 on Build GRCh38
rs745435492 Chr.5: 141489215-141489215 on Build GRCh38

View Full Product Details