Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30598713-30598982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GARS
Entrez Gene ID
2617
Gene Name
glycyl-tRNA synthetase
Gene Aliases
CMT2D, DSMAV, GlyRS, HMN5, SMAD1
UniGene
Hs.404321
Gene Chromosome Location
Chr.7: 30594735-30634033 on Build GRCh38
Transcripts
NM_001316772.1 NM_002047.3 XM_006715686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5013912 c.230T>G p.L77R Chr.7: 30598803-30598803 on Build GRCh38
485258 c.275A>G p.D92G Chr.7: 30598848-30598848 on Build GRCh38
3880552 c.236G>A p.R79Q Chr.7: 30598809-30598809 on Build GRCh38
185044 c.302G>A p.R101H Chr.7: 30598875-30598875 on Build GRCh38
4429850 c.300C>G p.A100A Chr.7: 30598873-30598873 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775444919 Chr.7: 30598812-30598812 on Build GRCh38
rs571004464 Chr.7: 30598855-30598855 on Build GRCh38
rs781432743 Chr.7: 30598873-30598873 on Build GRCh38
rs371564065 Chr.7: 30598781-30598781 on Build GRCh38
rs777889388 Chr.7: 30598769-30598769 on Build GRCh38
rs917554333 Chr.7: 30598791-30598791 on Build GRCh38
rs771656019 Chr.7: 30598918-30598918 on Build GRCh38
rs374616031 Chr.7: 30598844-30598844 on Build GRCh38
rs984100715 Chr.7: 30598831-30598831 on Build GRCh38
rs773748010 Chr.7: 30598892-30598892 on Build GRCh38
rs376199921 Chr.7: 30598807-30598807 on Build GRCh38
rs757026838 Chr.7: 30598920-30598922 on Build GRCh38
rs377155080 Chr.7: 30598948-30598948 on Build GRCh38
rs760384921 Chr.7: 30598924-30598924 on Build GRCh38
rs751597051 Chr.7: 30598856-30598856 on Build GRCh38
rs878874780 Chr.7: 30598743-30598743 on Build GRCh38
rs747257211 Chr.7: 30598771-30598771 on Build GRCh38
rs776884508 Chr.7: 30598837-30598837 on Build GRCh38
rs749199700 Chr.7: 30598824-30598824 on Build GRCh38
rs369898799 Chr.7: 30598843-30598843 on Build GRCh38
rs377122472 Chr.7: 30598827-30598827 on Build GRCh38
rs556718980 Chr.7: 30598926-30598926 on Build GRCh38
rs780858299 Chr.7: 30598800-30598800 on Build GRCh38
rs575420383 Chr.7: 30598945-30598945 on Build GRCh38
rs761378110 Chr.7: 30598833-30598833 on Build GRCh38
rs758964807 Chr.7: 30598845-30598845 on Build GRCh38
rs369466037 Chr.7: 30598809-30598809 on Build GRCh38
rs201728920 Chr.7: 30598835-30598835 on Build GRCh38
rs763563426 Chr.7: 30598930-30598930 on Build GRCh38
rs746056671 Chr.7: 30598874-30598874 on Build GRCh38
rs750700531 Chr.7: 30598949-30598949 on Build GRCh38
rs770063607 Chr.7: 30598956-30598956 on Build GRCh38
rs751337889 Chr.7: 30598878-30598879 on Build GRCh38
rs369681546 Chr.7: 30598742-30598742 on Build GRCh38
rs1030792388 Chr.7: 30598803-30598803 on Build GRCh38
rs372776194 Chr.7: 30598900-30598900 on Build GRCh38
rs768571328 Chr.7: 30598882-30598882 on Build GRCh38
rs757439624 Chr.7: 30598859-30598859 on Build GRCh38
rs751619983 Chr.7: 30598943-30598943 on Build GRCh38
rs750880484 Chr.7: 30598920-30598920 on Build GRCh38
rs1037845563 Chr.7: 30598897-30598897 on Build GRCh38
rs200887429 Chr.7: 30598875-30598875 on Build GRCh38
rs200294578 Chr.7: 30598826-30598826 on Build GRCh38
rs764610117 Chr.7: 30598932-30598932 on Build GRCh38
rs759941591 Chr.7: 30598842-30598842 on Build GRCh38
rs369224847 Chr.7: 30598878-30598878 on Build GRCh38
rs907969416 Chr.7: 30598940-30598940 on Build GRCh38
rs767369234 Chr.7: 30598923-30598924 on Build GRCh38

View Full Product Details