Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140807176-140807427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_031500.2

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_031500.2

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_031500.2

Gene Symbol
PCDHA4
Entrez Gene ID
56144
Gene Name
protocadherin alpha 4
Gene Aliases
CNR1, CNRN1, CRNR1, PCDH-ALPHA4
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140807074-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_031500.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3850837 c.38G>A p.R13H Chr.5: 140807225-140807225 on Build GRCh38
5425464 c.165G>T p.E55D Chr.5: 140807352-140807352 on Build GRCh38
5819763 c.224T>A p.L75Q Chr.5: 140807411-140807411 on Build GRCh38
4159467 c.212G>A p.R71H Chr.5: 140807399-140807399 on Build GRCh38
5425463 c.165G>T p.E55D Chr.5: 140807352-140807352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781892192 Chr.5: 140807325-140807325 on Build GRCh38
rs782668937 Chr.5: 140807390-140807390 on Build GRCh38
rs782322064 Chr.5: 140807401-140807401 on Build GRCh38
rs17844273 Chr.5: 140807405-140807405 on Build GRCh38
rs147378308 Chr.5: 140807215-140807215 on Build GRCh38
rs782077349 Chr.5: 140807404-140807404 on Build GRCh38
rs781836532 Chr.5: 140807250-140807250 on Build GRCh38
rs782751422 Chr.5: 140807206-140807206 on Build GRCh38
rs782485620 Chr.5: 140807334-140807334 on Build GRCh38
rs782350961 Chr.5: 140807405-140807406 on Build GRCh38
rs782310722 Chr.5: 140807279-140807279 on Build GRCh38
rs782421232 Chr.5: 140807209-140807209 on Build GRCh38
rs918123295 Chr.5: 140807354-140807354 on Build GRCh38
rs782357546 Chr.5: 140807369-140807369 on Build GRCh38
rs782807464 Chr.5: 140807234-140807234 on Build GRCh38
rs782359747 Chr.5: 140807280-140807280 on Build GRCh38
rs782331571 Chr.5: 140807285-140807285 on Build GRCh38
rs781918542 Chr.5: 140807390-140807391 on Build GRCh38
rs782612187 Chr.5: 140807400-140807401 on Build GRCh38
rs553258055 Chr.5: 140807376-140807376 on Build GRCh38
rs782522800 Chr.5: 140807326-140807326 on Build GRCh38
rs3822354 Chr.5: 140807395-140807395 on Build GRCh38
rs782583105 Chr.5: 140807251-140807251 on Build GRCh38
rs201303975 Chr.5: 140807404-140807404 on Build GRCh38
rs3822352 Chr.5: 140807399-140807399 on Build GRCh38
rs781811191 Chr.5: 140807389-140807389 on Build GRCh38
rs949440782 Chr.5: 140807357-140807357 on Build GRCh38
rs983753227 Chr.5: 140807303-140807303 on Build GRCh38
rs3822350 Chr.5: 140807403-140807403 on Build GRCh38
rs782067022 Chr.5: 140807391-140807392 on Build GRCh38
rs782764363 Chr.5: 140807395-140807396 on Build GRCh38
rs199939862 Chr.5: 140807327-140807327 on Build GRCh38
rs373851381 Chr.5: 140807346-140807346 on Build GRCh38
rs528454810 Chr.5: 140807296-140807296 on Build GRCh38
rs3822355 Chr.5: 140807394-140807394 on Build GRCh38
rs3822353 Chr.5: 140807397-140807397 on Build GRCh38
rs782494406 Chr.5: 140807387-140807387 on Build GRCh38
rs782426458 Chr.5: 140807260-140807260 on Build GRCh38
rs782255679 Chr.5: 140807391-140807391 on Build GRCh38
rs543774145 Chr.5: 140807400-140807400 on Build GRCh38
rs67934344 Chr.5: 140807394-140807394 on Build GRCh38
rs138132729 Chr.5: 140807259-140807259 on Build GRCh38
rs782029376 Chr.5: 140807288-140807288 on Build GRCh38
rs782815519 Chr.5: 140807315-140807315 on Build GRCh38
rs782241518 Chr.5: 140807368-140807368 on Build GRCh38
rs782084195 Chr.5: 140807283-140807283 on Build GRCh38
rs781879266 Chr.5: 140807237-140807237 on Build GRCh38
rs782639557 Chr.5: 140807248-140807248 on Build GRCh38
rs782704946 Chr.5: 140807377-140807377 on Build GRCh38
rs782794187 Chr.5: 140807393-140807394 on Build GRCh38
rs782371705 Chr.5: 140807403-140807403 on Build GRCh38
rs953996588 Chr.5: 140807223-140807223 on Build GRCh38
rs781978494 Chr.5: 140807275-140807275 on Build GRCh38
rs782375351 Chr.5: 140807273-140807273 on Build GRCh38
rs782782841 Chr.5: 140807383-140807383 on Build GRCh38
rs781951265 Chr.5: 140807373-140807373 on Build GRCh38
rs782301855 Chr.5: 140807203-140807204 on Build GRCh38
rs11167605 Chr.5: 140807352-140807352 on Build GRCh38
rs782005139 Chr.5: 140807396-140807396 on Build GRCh38
rs782120489 Chr.5: 140807398-140807398 on Build GRCh38
rs782505608 Chr.5: 140807239-140807239 on Build GRCh38
rs148298330 Chr.5: 140807381-140807381 on Build GRCh38
rs145024223 Chr.5: 140807362-140807362 on Build GRCh38
rs3822351 Chr.5: 140807401-140807401 on Build GRCh38
rs557517265 Chr.5: 140807268-140807268 on Build GRCh38
rs782721399 Chr.5: 140807306-140807306 on Build GRCh38
rs781857589 Chr.5: 140807386-140807386 on Build GRCh38
rs386692888 Chr.5: 140807403-140807406 on Build GRCh38
rs781800843 Chr.5: 140807311-140807311 on Build GRCh38
rs189135254 Chr.5: 140807302-140807302 on Build GRCh38
rs200172095 Chr.5: 140807396-140807397 on Build GRCh38

View Full Product Details