Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102022076-102022349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1R2
Entrez Gene ID
7850
Gene Name
interleukin 1 receptor type 2
Gene Aliases
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB
UniGene
Hs.25333
Gene Chromosome Location
Chr.2: 101991805-102028544 on Build GRCh38
Transcripts
NM_001261419.1 NM_004633.3 NR_048564.1 XM_006712734.3 XM_006712736.3 XM_011511801.2 XM_011511803.2 XM_011511804.2 XM_011511805.2 XM_011511808.2 XM_017004889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4128067 c.732G>C p.K244N Chr.2: 102022230-102022230 on Build GRCh38
2980444 c.732G>A p.K244K Chr.2: 102022230-102022230 on Build GRCh38
4799718 c.709C>A p.P237T Chr.2: 102022207-102022207 on Build GRCh38
3564408 c.726C>T p.P242P Chr.2: 102022224-102022224 on Build GRCh38
2980441 c.689delA p.E233fs*6 Chr.2: 102022187-102022187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948295855 Chr.2: 102022294-102022294 on Build GRCh38
rs778774050 Chr.2: 102022297-102022297 on Build GRCh38
rs3218959 Chr.2: 102022153-102022153 on Build GRCh38
rs370995625 Chr.2: 102022282-102022282 on Build GRCh38
rs369886122 Chr.2: 102022222-102022222 on Build GRCh38
rs1006077044 Chr.2: 102022113-102022113 on Build GRCh38
rs758651036 Chr.2: 102022223-102022223 on Build GRCh38
rs781078644 Chr.2: 102022287-102022287 on Build GRCh38
rs3218960 Chr.2: 102022171-102022171 on Build GRCh38
rs771416747 Chr.2: 102022259-102022259 on Build GRCh38
rs748873536 Chr.2: 102022216-102022217 on Build GRCh38
rs933762515 Chr.2: 102022270-102022270 on Build GRCh38
rs757509448 Chr.2: 102022285-102022285 on Build GRCh38
rs753294181 Chr.2: 102022277-102022277 on Build GRCh38
rs373531762 Chr.2: 102022224-102022224 on Build GRCh38
rs1047449220 Chr.2: 102022306-102022306 on Build GRCh38
rs765874077 Chr.2: 102022274-102022274 on Build GRCh38
rs187608088 Chr.2: 102022279-102022279 on Build GRCh38
rs3218961 Chr.2: 102022183-102022183 on Build GRCh38
rs183091210 Chr.2: 102022236-102022236 on Build GRCh38
rs768536544 Chr.2: 102022241-102022241 on Build GRCh38
rs192467108 Chr.2: 102022291-102022291 on Build GRCh38
rs374927636 Chr.2: 102022137-102022137 on Build GRCh38
rs771086408 Chr.2: 102022149-102022149 on Build GRCh38
rs1003457521 Chr.2: 102022315-102022315 on Build GRCh38
rs199970902 Chr.2: 102022244-102022244 on Build GRCh38
rs773839071 Chr.2: 102022285-102022286 on Build GRCh38
rs771844597 Chr.2: 102022284-102022284 on Build GRCh38
rs754295491 Chr.2: 102022202-102022202 on Build GRCh38
rs907629287 Chr.2: 102022307-102022307 on Build GRCh38
rs749273300 Chr.2: 102022293-102022293 on Build GRCh38
rs1031510163 Chr.2: 102022320-102022320 on Build GRCh38
rs749113919 Chr.2: 102022238-102022238 on Build GRCh38
rs375661356 Chr.2: 102022234-102022234 on Build GRCh38
rs267598801 Chr.2: 102022215-102022215 on Build GRCh38
rs775265303 Chr.2: 102022165-102022165 on Build GRCh38
rs941828992 Chr.2: 102022309-102022309 on Build GRCh38
rs956746970 Chr.2: 102022176-102022176 on Build GRCh38
rs1037561257 Chr.2: 102022326-102022326 on Build GRCh38
rs996159387 Chr.2: 102022132-102022132 on Build GRCh38
rs770584494 Chr.2: 102022219-102022220 on Build GRCh38
rs773871026 Chr.2: 102022179-102022179 on Build GRCh38
rs1047943600 Chr.2: 102022100-102022100 on Build GRCh38
rs745859334 Chr.2: 102022157-102022157 on Build GRCh38
rs762532597 Chr.2: 102022169-102022169 on Build GRCh38
rs373351875 Chr.2: 102022268-102022268 on Build GRCh38
rs772911553 Chr.2: 102022186-102022187 on Build GRCh38
rs886234783 Chr.2: 102022105-102022105 on Build GRCh38
rs369134425 Chr.2: 102022225-102022225 on Build GRCh38
rs747093682 Chr.2: 102022147-102022147 on Build GRCh38
rs548265414 Chr.2: 102022329-102022329 on Build GRCh38
rs748314810 Chr.2: 102022136-102022136 on Build GRCh38
rs760070029 Chr.2: 102022271-102022271 on Build GRCh38
rs768322521 Chr.2: 102022295-102022295 on Build GRCh38
rs900448658 Chr.2: 102022128-102022128 on Build GRCh38
rs368745591 Chr.2: 102022158-102022158 on Build GRCh38
rs763383186 Chr.2: 102022278-102022278 on Build GRCh38
rs563001194 Chr.2: 102022289-102022289 on Build GRCh38
rs761592613 Chr.2: 102022254-102022254 on Build GRCh38
rs1037593746 Chr.2: 102022119-102022119 on Build GRCh38
rs958988768 Chr.2: 102022240-102022240 on Build GRCh38

View Full Product Details