Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 46549574-46549303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRIN2
Entrez Gene ID
9721
Gene Name
G protein regulated inducer of neurite outgrowth 2
Gene Aliases
GRIN2, KIAA0514
UniGene
Hs.523375
Gene Chromosome Location
Chr.10: 46543963-46556637 on Build GRCh38
Transcripts
NM_014696.3 XM_005277666.3 XM_011540388.2 XM_011540395.2 XM_011540400.2 XM_017016974.1 XM_017016975.1 XM_017016976.1 XM_017016977.1 XM_017016978.1 XM_017016979.1 XM_017016980.1 XM_017016981.1 XM_017016982.1 XM_017016983.1 XM_017016984.1 XM_017017029.1 XM_017017030.1 XM_017017031.1 XM_017017032.1 XM_017017033.1 XM_017017034.1 XM_017017038.1 XM_017017039.1 XM_017017040.1

Gene Symbol
LOC107984026
Entrez Gene ID
107984026
Gene Name
arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.10: 46335552-46838268 on Build GRCh38
Transcripts
NM_014696.3 XM_005277666.3 XM_011540388.2 XM_011540395.2 XM_011540400.2 XM_017016974.1 XM_017016975.1 XM_017016976.1 XM_017016977.1 XM_017016978.1 XM_017016979.1 XM_017016980.1 XM_017016981.1 XM_017016982.1 XM_017016983.1 XM_017016984.1 XM_017017029.1 XM_017017030.1 XM_017017031.1 XM_017017032.1 XM_017017033.1 XM_017017034.1 XM_017017038.1 XM_017017039.1 XM_017017040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4014310 c.1182C>T p.Y394Y Chr.10: 46549555-46549555 on Build GRCh38
255923 c.1336C>T p.R446C Chr.10: 46549401-46549401 on Build GRCh38
4894477 c.1330C>T p.L444L Chr.10: 46549407-46549407 on Build GRCh38
3751795 c.1308A>G p.P436P Chr.10: 46549429-46549429 on Build GRCh38
4144708 c.1199T>C p.L400P Chr.10: 46549538-46549538 on Build GRCh38
View More

View Full Product Details