Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 547351-547590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZMM
Entrez Gene ID
3004
Gene Name
granzyme M
Gene Aliases
LMET1, MET1
UniGene
Hs.465511
Gene Chromosome Location
Chr.19: 544034-549920 on Build GRCh38
Transcripts
NM_001258351.1 NM_005317.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6219969 c.175A>G p.K59E Chr.19: 547399-547399 on Build GRCh38
7290597 c.211C>T p.R71W Chr.19: 547435-547435 on Build GRCh38
4689402 c.189G>A p.T63T Chr.19: 547413-547413 on Build GRCh38
2929880 c.154G>A p.G52R Chr.19: 547378-547378 on Build GRCh38
3960453 c.154G>T p.G52W Chr.19: 547378-547378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942980035 Chr.19: 547476-547476 on Build GRCh38
rs751910126 Chr.19: 547427-547427 on Build GRCh38
rs764766156 Chr.19: 547488-547488 on Build GRCh38
rs769969742 Chr.19: 547377-547377 on Build GRCh38
rs780211377 Chr.19: 547448-547448 on Build GRCh38
rs779153027 Chr.19: 547449-547458 on Build GRCh38
rs757450436 Chr.19: 547370-547370 on Build GRCh38
rs185429686 Chr.19: 547544-547544 on Build GRCh38
rs759077712 Chr.19: 547485-547485 on Build GRCh38
rs2070796 Chr.19: 547555-547555 on Build GRCh38
rs780087868 Chr.19: 547406-547415 on Build GRCh38
rs755067260 Chr.19: 547415-547416 on Build GRCh38
rs146522321 Chr.19: 547378-547378 on Build GRCh38
rs374939287 Chr.19: 547412-547412 on Build GRCh38
rs992020751 Chr.19: 547516-547516 on Build GRCh38
rs543874497 Chr.19: 547404-547404 on Build GRCh38
rs746140221 Chr.19: 547376-547376 on Build GRCh38
rs774142092 Chr.19: 547390-547390 on Build GRCh38
rs527907006 Chr.19: 547500-547500 on Build GRCh38
rs1051454332 Chr.19: 547492-547492 on Build GRCh38
rs761539542 Chr.19: 547401-547401 on Build GRCh38
rs756467224 Chr.19: 547438-547438 on Build GRCh38
rs768715164 Chr.19: 547380-547380 on Build GRCh38
rs767160387 Chr.19: 547402-547402 on Build GRCh38
rs768668456 Chr.19: 547460-547460 on Build GRCh38
rs770576095 Chr.19: 547482-547482 on Build GRCh38
rs753936908 Chr.19: 547410-547411 on Build GRCh38
rs2070795 Chr.19: 547491-547491 on Build GRCh38
rs948004680 Chr.19: 547540-547540 on Build GRCh38
rs1034717815 Chr.19: 547560-547560 on Build GRCh38
rs908920474 Chr.19: 547545-547545 on Build GRCh38
rs764607104 Chr.19: 547426-547426 on Build GRCh38
rs746061534 Chr.19: 547436-547436 on Build GRCh38
rs199725777 Chr.19: 547467-547467 on Build GRCh38
rs765862575 Chr.19: 547413-547413 on Build GRCh38
rs781655159 Chr.19: 547431-547431 on Build GRCh38
rs749396827 Chr.19: 547458-547458 on Build GRCh38
rs140327613 Chr.19: 547372-547372 on Build GRCh38
rs753412913 Chr.19: 547416-547416 on Build GRCh38
rs755991300 Chr.19: 547405-547406 on Build GRCh38
rs538294785 Chr.19: 547554-547554 on Build GRCh38
rs193277130 Chr.19: 547532-547532 on Build GRCh38
rs778751059 Chr.19: 547465-547465 on Build GRCh38
rs147907546 Chr.19: 547429-547429 on Build GRCh38
rs773118602 Chr.19: 547470-547470 on Build GRCh38
rs760316637 Chr.19: 547473-547473 on Build GRCh38
rs369284240 Chr.19: 547469-547469 on Build GRCh38
rs2070797 Chr.19: 547561-547561 on Build GRCh38
rs2070794 Chr.19: 547483-547483 on Build GRCh38
rs933455848 Chr.19: 547569-547569 on Build GRCh38

View Full Product Details