Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17616480-17616741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP153
Entrez Gene ID
9972
Gene Name
nucleoporin 153
Gene Aliases
HNUP153, N153
UniGene
Hs.601591
Gene Chromosome Location
Chr.6: 17615035-17706834 on Build GRCh38
Transcripts
NM_001278209.1 NM_001278210.1 NM_005124.3 XM_005249507.2 XM_006715290.2 XM_011515028.2 XM_017011594.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
283430 c.4298C>T p.S1433L Chr.6: 17616572-17616572 on Build GRCh38
5688572 c.4351A>T p.N1451Y Chr.6: 17616612-17616612 on Build GRCh38
4646772 c.4237G>A p.G1413R Chr.6: 17616633-17616633 on Build GRCh38
5869814 c.4369G>A p.A1457T Chr.6: 17616594-17616594 on Build GRCh38
5567394 c.4178C>T p.S1393L Chr.6: 17616692-17616692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555051041 Chr.6: 17616515-17616515 on Build GRCh38
rs202228382 Chr.6: 17616631-17616631 on Build GRCh38
rs563893774 Chr.6: 17616571-17616571 on Build GRCh38
rs767521864 Chr.6: 17616578-17616578 on Build GRCh38
rs200059221 Chr.6: 17616572-17616572 on Build GRCh38
rs748541496 Chr.6: 17616647-17616647 on Build GRCh38
rs1010809297 Chr.6: 17616573-17616573 on Build GRCh38
rs983084181 Chr.6: 17616673-17616673 on Build GRCh38
rs149664123 Chr.6: 17616652-17616652 on Build GRCh38
rs369751640 Chr.6: 17616678-17616678 on Build GRCh38
rs759121267 Chr.6: 17616520-17616520 on Build GRCh38
rs1014166852 Chr.6: 17616528-17616528 on Build GRCh38
rs749372288 Chr.6: 17616617-17616617 on Build GRCh38
rs780052978 Chr.6: 17616608-17616608 on Build GRCh38
rs1034741775 Chr.6: 17616602-17616602 on Build GRCh38
rs763785323 Chr.6: 17616684-17616684 on Build GRCh38
rs764588482 Chr.6: 17616521-17616521 on Build GRCh38
rs771458324 Chr.6: 17616663-17616663 on Build GRCh38
rs1034984681 Chr.6: 17616513-17616513 on Build GRCh38
rs757863134 Chr.6: 17616546-17616546 on Build GRCh38
rs115920718 Chr.6: 17616702-17616702 on Build GRCh38
rs774932372 Chr.6: 17616666-17616666 on Build GRCh38
rs747419051 Chr.6: 17616661-17616661 on Build GRCh38
rs541329586 Chr.6: 17616547-17616547 on Build GRCh38
rs969173830 Chr.6: 17616584-17616584 on Build GRCh38
rs199739841 Chr.6: 17616560-17616560 on Build GRCh38
rs755062586 Chr.6: 17616565-17616566 on Build GRCh38
rs951180822 Chr.6: 17616615-17616615 on Build GRCh38
rs973622722 Chr.6: 17616671-17616671 on Build GRCh38
rs201853569 Chr.6: 17616567-17616567 on Build GRCh38
rs773777225 Chr.6: 17616656-17616656 on Build GRCh38
rs140496420 Chr.6: 17616695-17616695 on Build GRCh38
rs748383680 Chr.6: 17616700-17616700 on Build GRCh38
rs372772287 Chr.6: 17616691-17616691 on Build GRCh38
rs779251127 Chr.6: 17616636-17616636 on Build GRCh38
rs778950995 Chr.6: 17616642-17616644 on Build GRCh38
rs761600273 Chr.6: 17616692-17616692 on Build GRCh38
rs753704784 Chr.6: 17616511-17616512 on Build GRCh38
rs772751209 Chr.6: 17616509-17616509 on Build GRCh38
rs142342732 Chr.6: 17616586-17616586 on Build GRCh38
rs149096395 Chr.6: 17616534-17616534 on Build GRCh38
rs756071154 Chr.6: 17616711-17616711 on Build GRCh38
rs756230394 Chr.6: 17616598-17616598 on Build GRCh38
rs775224494 Chr.6: 17616531-17616531 on Build GRCh38

View Full Product Details