Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64986905-64987166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3

Gene Symbol
FNTB
Entrez Gene ID
2342
Gene Name
farnesyltransferase, CAAX box, beta
Gene Aliases
FPTB
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64986789-65062652 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4436761 c.51C>T p.P17P Chr.14: 64987004-64987004 on Build GRCh38
2248800 c.131C>A p.T44K Chr.14: 64987084-64987084 on Build GRCh38
344312 c.41C>G p.S14C Chr.14: 64986994-64986994 on Build GRCh38
6218616 c.122_124delAAA p.E41_T42>A Chr.14: 64987075-64987077 on Build GRCh38
4672416 c.116C>A p.S39* Chr.14: 64987069-64987069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778887316 Chr.14: 64986970-64986970 on Build GRCh38
rs141330083 Chr.14: 64986989-64986989 on Build GRCh38
rs373810742 Chr.14: 64986937-64986937 on Build GRCh38
rs371704533 Chr.14: 64986949-64986949 on Build GRCh38
rs561584193 Chr.14: 64986985-64986985 on Build GRCh38
rs751307216 Chr.14: 64987143-64987143 on Build GRCh38
rs539673690 Chr.14: 64986932-64986932 on Build GRCh38
rs780965060 Chr.14: 64987128-64987128 on Build GRCh38
rs191661409 Chr.14: 64987138-64987138 on Build GRCh38
rs759929939 Chr.14: 64987091-64987091 on Build GRCh38
rs747665462 Chr.14: 64987148-64987148 on Build GRCh38
rs754558517 Chr.14: 64987108-64987109 on Build GRCh38
rs780651873 Chr.14: 64987147-64987147 on Build GRCh38
rs747645178 Chr.14: 64987085-64987085 on Build GRCh38
rs375130871 Chr.14: 64987108-64987108 on Build GRCh38
rs774776136 Chr.14: 64987090-64987090 on Build GRCh38
rs757328464 Chr.14: 64987046-64987046 on Build GRCh38
rs890782060 Chr.14: 64987145-64987145 on Build GRCh38
rs943217761 Chr.14: 64987126-64987126 on Build GRCh38
rs768162893 Chr.14: 64987067-64987067 on Build GRCh38
rs754719425 Chr.14: 64987146-64987146 on Build GRCh38
rs368373272 Chr.14: 64987130-64987130 on Build GRCh38
rs746503324 Chr.14: 64986980-64986980 on Build GRCh38
rs749196775 Chr.14: 64986965-64986965 on Build GRCh38
rs757093033 Chr.14: 64986933-64986936 on Build GRCh38
rs771908354 Chr.14: 64986977-64986977 on Build GRCh38
rs752927586 Chr.14: 64987005-64987005 on Build GRCh38
rs774755912 Chr.14: 64986986-64986986 on Build GRCh38
rs374564162 Chr.14: 64987013-64987013 on Build GRCh38
rs768170684 Chr.14: 64986981-64986981 on Build GRCh38
rs761924634 Chr.14: 64987125-64987125 on Build GRCh38
rs778979315 Chr.14: 64987049-64987049 on Build GRCh38
rs779602842 Chr.14: 64987054-64987054 on Build GRCh38
rs777605646 Chr.14: 64986963-64986963 on Build GRCh38
rs769518122 Chr.14: 64987001-64987001 on Build GRCh38
rs758200053 Chr.14: 64987053-64987053 on Build GRCh38
rs779939946 Chr.14: 64987139-64987139 on Build GRCh38
rs908171354 Chr.14: 64986992-64986992 on Build GRCh38
rs1047426368 Chr.14: 64987149-64987149 on Build GRCh38
rs775983734 Chr.14: 64986982-64986982 on Build GRCh38
rs764279329 Chr.14: 64987019-64987019 on Build GRCh38
rs767735610 Chr.14: 64987004-64987004 on Build GRCh38
rs959730328 Chr.14: 64986947-64986947 on Build GRCh38
rs372708444 Chr.14: 64986930-64986930 on Build GRCh38
rs145259305 Chr.14: 64987025-64987025 on Build GRCh38
rs541920896 Chr.14: 64986968-64986968 on Build GRCh38
rs750346128 Chr.14: 64987133-64987133 on Build GRCh38
rs200225795 Chr.14: 64986962-64986962 on Build GRCh38
rs147689181 Chr.14: 64987050-64987050 on Build GRCh38
rs932410042 Chr.14: 64987131-64987131 on Build GRCh38
rs866620194 Chr.14: 64987041-64987041 on Build GRCh38
rs746724957 Chr.14: 64987065-64987065 on Build GRCh38
rs777849808 Chr.14: 64987140-64987140 on Build GRCh38
rs766780991 Chr.14: 64986935-64986935 on Build GRCh38
rs765673443 Chr.14: 64986929-64986929 on Build GRCh38
rs767823775 Chr.14: 64987092-64987092 on Build GRCh38
rs755260685 Chr.14: 64986938-64986938 on Build GRCh38
rs775150635 Chr.14: 64986979-64986979 on Build GRCh38
rs575336097 Chr.14: 64987120-64987120 on Build GRCh38
rs754004557 Chr.14: 64987122-64987122 on Build GRCh38
rs745650313 Chr.14: 64986976-64986976 on Build GRCh38
rs763250558 Chr.14: 64986933-64986933 on Build GRCh38
rs771444910 Chr.14: 64987089-64987089 on Build GRCh38
rs200261877 Chr.14: 64986948-64986948 on Build GRCh38
rs919645186 Chr.14: 64987077-64987077 on Build GRCh38
rs553529626 Chr.14: 64986941-64986941 on Build GRCh38
rs776273553 Chr.14: 64987074-64987074 on Build GRCh38
rs756154826 Chr.14: 64986961-64986961 on Build GRCh38
rs960247469 Chr.14: 64987017-64987017 on Build GRCh38
rs760993482 Chr.14: 64987116-64987116 on Build GRCh38

View Full Product Details