Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 36991526-36991785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPM2AIP1
Entrez Gene ID
9852
Gene Name
EPM2A interacting protein 1
Gene Aliases
-
UniGene
Hs.28020
Gene Chromosome Location
Chr.3: 36985866-36993304 on Build GRCh38
Transcripts
NM_014805.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3427533 c.1412T>C p.F471S Chr.3: 36991666-36991666 on Build GRCh38
5438725 c.1516G>A p.E506K Chr.3: 36991562-36991562 on Build GRCh38
5994534 c.1531C>T p.Q511* Chr.3: 36991547-36991547 on Build GRCh38
4983996 c.1528C>G p.L510V Chr.3: 36991550-36991550 on Build GRCh38
730241 c.1351G>C p.D451H Chr.3: 36991727-36991727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761152116 Chr.3: 36991749-36991749 on Build GRCh38
rs556246683 Chr.3: 36991757-36991757 on Build GRCh38
rs760126765 Chr.3: 36991725-36991725 on Build GRCh38
rs946489432 Chr.3: 36991673-36991673 on Build GRCh38
rs774713918 Chr.3: 36991715-36991715 on Build GRCh38
rs781091412 Chr.3: 36991683-36991683 on Build GRCh38
rs34013758 Chr.3: 36991635-36991635 on Build GRCh38
rs769400518 Chr.3: 36991692-36991692 on Build GRCh38
rs761992366 Chr.3: 36991658-36991658 on Build GRCh38
rs779795438 Chr.3: 36991656-36991656 on Build GRCh38
rs754125935 Chr.3: 36991760-36991760 on Build GRCh38
rs768027698 Chr.3: 36991647-36991647 on Build GRCh38
rs368540000 Chr.3: 36991726-36991726 on Build GRCh38
rs756450483 Chr.3: 36991584-36991584 on Build GRCh38
rs374061153 Chr.3: 36991565-36991565 on Build GRCh38
rs778163588 Chr.3: 36991589-36991589 on Build GRCh38
rs762395461 Chr.3: 36991708-36991708 on Build GRCh38
rs753053880 Chr.3: 36991745-36991745 on Build GRCh38
rs747959787 Chr.3: 36991686-36991686 on Build GRCh38
rs768306003 Chr.3: 36991681-36991681 on Build GRCh38
rs1000009385 Chr.3: 36991731-36991731 on Build GRCh38
rs749339315 Chr.3: 36991617-36991617 on Build GRCh38
rs868284480 Chr.3: 36991736-36991736 on Build GRCh38
rs375933286 Chr.3: 36991701-36991701 on Build GRCh38
rs937530591 Chr.3: 36991633-36991633 on Build GRCh38
rs370427836 Chr.3: 36991658-36991658 on Build GRCh38
rs748843893 Chr.3: 36991605-36991605 on Build GRCh38

View Full Product Details