Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32088455-32088724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCK
Entrez Gene ID
3055
Gene Name
HCK proto-oncogene, Src family tyrosine kinase
Gene Aliases
JTK9, p59Hck, p61Hck
UniGene
Hs.655210
Gene Chromosome Location
Chr.20: 32052188-32101856 on Build GRCh38
Transcripts
NM_001172129.1 NM_001172130.1 NM_001172131.1 NM_001172132.1 NM_001172133.1 NM_002110.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1025673 c.1011C>T p.L337L Chr.20: 32088626-32088626 on Build GRCh38
577192 c.1016G>A p.G339E Chr.20: 32088568-32088568 on Build GRCh38
1025671 c.1074C>T p.L358L Chr.20: 32088626-32088626 on Build GRCh38
6677980 c.1023C>A p.F341L Chr.20: 32088635-32088635 on Build GRCh38
2889003 c.1035G>A p.L345L Chr.20: 32088587-32088587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939529459 Chr.20: 32088602-32088602 on Build GRCh38
rs2223785 Chr.20: 32088513-32088513 on Build GRCh38
rs577270836 Chr.20: 32088654-32088654 on Build GRCh38
rs137871550 Chr.20: 32088564-32088564 on Build GRCh38
rs750744039 Chr.20: 32088683-32088683 on Build GRCh38
rs778936221 Chr.20: 32088521-32088525 on Build GRCh38
rs758555670 Chr.20: 32088696-32088696 on Build GRCh38
rs752703448 Chr.20: 32088545-32088545 on Build GRCh38
rs771849634 Chr.20: 32088605-32088605 on Build GRCh38
rs199570263 Chr.20: 32088657-32088657 on Build GRCh38
rs746617419 Chr.20: 32088688-32088689 on Build GRCh38
rs765791760 Chr.20: 32088521-32088521 on Build GRCh38
rs973410419 Chr.20: 32088538-32088538 on Build GRCh38
rs182955378 Chr.20: 32088658-32088658 on Build GRCh38
rs755187545 Chr.20: 32088586-32088586 on Build GRCh38
rs779172321 Chr.20: 32088689-32088689 on Build GRCh38
rs759520283 Chr.20: 32088562-32088562 on Build GRCh38
rs770671364 Chr.20: 32088689-32088689 on Build GRCh38
rs954670632 Chr.20: 32088503-32088503 on Build GRCh38
rs567235368 Chr.20: 32088547-32088547 on Build GRCh38
rs757678615 Chr.20: 32088556-32088556 on Build GRCh38
rs768182335 Chr.20: 32088557-32088557 on Build GRCh38
rs767983967 Chr.20: 32088677-32088677 on Build GRCh38
rs977534723 Chr.20: 32088493-32088493 on Build GRCh38
rs749323511 Chr.20: 32088572-32088572 on Build GRCh38
rs757805734 Chr.20: 32088562-32088563 on Build GRCh38
rs779059256 Chr.20: 32088599-32088599 on Build GRCh38
rs201979534 Chr.20: 32088561-32088561 on Build GRCh38
rs776537763 Chr.20: 32088498-32088498 on Build GRCh38
rs1037971062 Chr.20: 32088653-32088653 on Build GRCh38
rs753328251 Chr.20: 32088522-32088522 on Build GRCh38
rs369207649 Chr.20: 32088559-32088559 on Build GRCh38
rs748354614 Chr.20: 32088695-32088695 on Build GRCh38
rs560990543 Chr.20: 32088671-32088671 on Build GRCh38
rs74654967 Chr.20: 32088666-32088666 on Build GRCh38
rs151054795 Chr.20: 32088628-32088628 on Build GRCh38
rs773190531 Chr.20: 32088621-32088621 on Build GRCh38
rs747978102 Chr.20: 32088601-32088601 on Build GRCh38
rs762339235 Chr.20: 32088668-32088668 on Build GRCh38
rs554987957 Chr.20: 32088543-32088543 on Build GRCh38
rs369945538 Chr.20: 32088665-32088665 on Build GRCh38
rs751990347 Chr.20: 32088552-32088552 on Build GRCh38
rs531739593 Chr.20: 32088687-32088687 on Build GRCh38
rs777381549 Chr.20: 32088654-32088655 on Build GRCh38
rs764256093 Chr.20: 32088686-32088686 on Build GRCh38
rs987249636 Chr.20: 32088512-32088512 on Build GRCh38

View Full Product Details