Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62760085-62760338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTSR1
Entrez Gene ID
4923
Gene Name
neurotensin receptor 1 (high affinity)
Gene Aliases
NTR
UniGene
Hs.590869
Gene Chromosome Location
Chr.20: 62708770-62762771 on Build GRCh38
Transcripts
NM_002531.2 XM_011528827.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5604284 c.1087C>T p.L363L Chr.20: 62760097-62760097 on Build GRCh38
4564440 c.1194_1195GG>AA p.A399T Chr.20: 62760204-62760205 on Build GRCh38
3548741 c.1240C>T p.R414C Chr.20: 62760250-62760250 on Build GRCh38
7380636 c.1252T>C p.Y418H Chr.20: 62760262-62760262 on Build GRCh38
3548740 c.1169G>C p.R390T Chr.20: 62760179-62760179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756303943 Chr.20: 62760285-62760285 on Build GRCh38
rs771598068 Chr.20: 62760318-62760318 on Build GRCh38
rs148516370 Chr.20: 62760173-62760173 on Build GRCh38
rs373370662 Chr.20: 62760146-62760146 on Build GRCh38
rs3887638 Chr.20: 62760236-62760236 on Build GRCh38
rs149615776 Chr.20: 62760124-62760124 on Build GRCh38
rs535027791 Chr.20: 62760159-62760159 on Build GRCh38
rs780291520 Chr.20: 62760209-62760209 on Build GRCh38
rs373509518 Chr.20: 62760222-62760222 on Build GRCh38
rs140834874 Chr.20: 62760169-62760169 on Build GRCh38
rs754560333 Chr.20: 62760120-62760120 on Build GRCh38
rs6062580 Chr.20: 62760267-62760267 on Build GRCh38
rs552846115 Chr.20: 62760136-62760136 on Build GRCh38
rs372880112 Chr.20: 62760279-62760279 on Build GRCh38
rs749726085 Chr.20: 62760300-62760300 on Build GRCh38
rs140803345 Chr.20: 62760198-62760198 on Build GRCh38
rs765864962 Chr.20: 62760109-62760109 on Build GRCh38
rs142679048 Chr.20: 62760195-62760195 on Build GRCh38
rs45613333 Chr.20: 62760162-62760162 on Build GRCh38
rs878999120 Chr.20: 62760207-62760207 on Build GRCh38
rs948015881 Chr.20: 62760191-62760191 on Build GRCh38
rs771252845 Chr.20: 62760305-62760305 on Build GRCh38
rs202014500 Chr.20: 62760127-62760127 on Build GRCh38
rs200413077 Chr.20: 62760250-62760250 on Build GRCh38
rs771483855 Chr.20: 62760230-62760230 on Build GRCh38
rs16983222 Chr.20: 62760276-62760276 on Build GRCh38
rs752314674 Chr.20: 62760183-62760183 on Build GRCh38
rs755135046 Chr.20: 62760277-62760277 on Build GRCh38
rs751244883 Chr.20: 62760114-62760114 on Build GRCh38
rs780238434 Chr.20: 62760137-62760137 on Build GRCh38
rs749774646 Chr.20: 62760224-62760224 on Build GRCh38
rs747748048 Chr.20: 62760108-62760109 on Build GRCh38
rs144593693 Chr.20: 62760170-62760170 on Build GRCh38
rs774833545 Chr.20: 62760248-62760248 on Build GRCh38
rs775142005 Chr.20: 62760258-62760258 on Build GRCh38
rs746324764 Chr.20: 62760315-62760315 on Build GRCh38
rs747155473 Chr.20: 62760211-62760211 on Build GRCh38
rs143383902 Chr.20: 62760213-62760213 on Build GRCh38
rs766991609 Chr.20: 62760251-62760251 on Build GRCh38
rs770176811 Chr.20: 62760160-62760160 on Build GRCh38
rs557535083 Chr.20: 62760214-62760214 on Build GRCh38
rs753609077 Chr.20: 62760274-62760274 on Build GRCh38
rs199957018 Chr.20: 62760161-62760161 on Build GRCh38
rs773626025 Chr.20: 62760140-62760140 on Build GRCh38
rs779443873 Chr.20: 62760310-62760310 on Build GRCh38
rs773560374 Chr.20: 62760223-62760223 on Build GRCh38
rs533071110 Chr.20: 62760321-62760321 on Build GRCh38
rs142268215 Chr.20: 62760249-62760249 on Build GRCh38
rs772626247 Chr.20: 62760128-62760134 on Build GRCh38
rs767312330 Chr.20: 62760121-62760121 on Build GRCh38
rs748356190 Chr.20: 62760220-62760220 on Build GRCh38
rs747514690 Chr.20: 62760271-62760271 on Build GRCh38
rs376745440 Chr.20: 62760271-62760271 on Build GRCh38
rs202170385 Chr.20: 62760145-62760145 on Build GRCh38
rs201891030 Chr.20: 62760289-62760289 on Build GRCh38
rs761511271 Chr.20: 62760275-62760275 on Build GRCh38
rs370016377 Chr.20: 62760208-62760208 on Build GRCh38
rs747235475 Chr.20: 62760142-62760142 on Build GRCh38
rs73918681 Chr.20: 62760172-62760172 on Build GRCh38
rs151022006 Chr.20: 62760197-62760197 on Build GRCh38
rs967388196 Chr.20: 62760119-62760119 on Build GRCh38
rs377711186 Chr.20: 62760125-62760125 on Build GRCh38
rs73918683 Chr.20: 62760252-62760252 on Build GRCh38
rs201168089 Chr.20: 62760234-62760234 on Build GRCh38
rs762422861 Chr.20: 62760108-62760108 on Build GRCh38
rs74420799 Chr.20: 62760280-62760280 on Build GRCh38

View Full Product Details