Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95040509-95040760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2C1
Entrez Gene ID
7181
Gene Name
nuclear receptor subfamily 2 group C member 1
Gene Aliases
TR2
UniGene
Hs.108301
Gene Chromosome Location
Chr.12: 95020229-95073628 on Build GRCh38
Transcripts
NM_001032287.2 NM_001127362.1 NM_003297.3 XM_005269133.2 XM_006719584.2 XM_006719585.2 XM_011538716.2 XM_011538717.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4819004 c.1156G>A p.E386K Chr.12: 95040573-95040573 on Build GRCh38
4459605 c.1132C>T p.L378F Chr.12: 95040597-95040597 on Build GRCh38
6258476 c.1161C>A p.Y387* Chr.12: 95040568-95040568 on Build GRCh38
1299976 c.1203G>T p.L401L Chr.12: 95040526-95040526 on Build GRCh38
4811794 c.1203G>T p.L401L Chr.12: 95040526-95040526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770176318 Chr.12: 95040642-95040642 on Build GRCh38
rs779561570 Chr.12: 95040611-95040611 on Build GRCh38
rs369909050 Chr.12: 95040672-95040672 on Build GRCh38
rs183766995 Chr.12: 95040626-95040626 on Build GRCh38
rs758282593 Chr.12: 95040604-95040604 on Build GRCh38
rs750157020 Chr.12: 95040602-95040602 on Build GRCh38
rs914877300 Chr.12: 95040654-95040654 on Build GRCh38
rs74588396 Chr.12: 95040677-95040677 on Build GRCh38
rs750914239 Chr.12: 95040544-95040544 on Build GRCh38
rs747679832 Chr.12: 95040636-95040636 on Build GRCh38
rs780640517 Chr.12: 95040630-95040630 on Build GRCh38
rs11107870 Chr.12: 95040676-95040676 on Build GRCh38
rs189332543 Chr.12: 95040628-95040628 on Build GRCh38
rs759671166 Chr.12: 95040551-95040551 on Build GRCh38
rs949416304 Chr.12: 95040662-95040662 on Build GRCh38
rs778962119 Chr.12: 95040588-95040588 on Build GRCh38
rs990524007 Chr.12: 95040638-95040638 on Build GRCh38
rs757216462 Chr.12: 95040583-95040583 on Build GRCh38
rs752859112 Chr.12: 95040579-95040579 on Build GRCh38
rs773807849 Chr.12: 95040646-95040646 on Build GRCh38
rs767642382 Chr.12: 95040559-95040559 on Build GRCh38

View Full Product Details