Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121846705-121846940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPD
Entrez Gene ID
3242
Gene Name
4-hydroxyphenylpyruvate dioxygenase
Gene Aliases
4-HPPD, 4HPPD, GLOD3, HPPDASE, PPD
UniGene
Hs.2899
Gene Chromosome Location
Chr.12: 121839527-121888611 on Build GRCh38
Transcripts
NM_001171993.1 NM_002150.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6303159 c.830C>A p.A277E Chr.12: 121846863-121846863 on Build GRCh38
1638224 c.831G>A p.A277A Chr.12: 121846862-121846862 on Build GRCh38
3457218 c.760G>A p.E254K Chr.12: 121846933-121846933 on Build GRCh38
1359522 c.822C>A p.I274I Chr.12: 121846871-121846871 on Build GRCh38
6997133 c.783C>T p.G261G Chr.12: 121846910-121846910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032751884 Chr.12: 121846754-121846754 on Build GRCh38
rs550194762 Chr.12: 121846810-121846810 on Build GRCh38
rs774349655 Chr.12: 121846851-121846851 on Build GRCh38
rs529795459 Chr.12: 121846879-121846879 on Build GRCh38
rs768597159 Chr.12: 121846846-121846846 on Build GRCh38
rs183844361 Chr.12: 121846840-121846840 on Build GRCh38
rs773284217 Chr.12: 121846870-121846870 on Build GRCh38
rs981725516 Chr.12: 121846788-121846788 on Build GRCh38
rs753320389 Chr.12: 121846902-121846902 on Build GRCh38
rs2247291 Chr.12: 121846809-121846809 on Build GRCh38
rs974374228 Chr.12: 121846873-121846873 on Build GRCh38
rs201249721 Chr.12: 121846916-121846916 on Build GRCh38
rs915720759 Chr.12: 121846781-121846781 on Build GRCh38
rs1030101490 Chr.12: 121846786-121846786 on Build GRCh38
rs769448651 Chr.12: 121846831-121846831 on Build GRCh38
rs765840049 Chr.12: 121846896-121846896 on Build GRCh38
rs564460302 Chr.12: 121846766-121846766 on Build GRCh38
rs971348200 Chr.12: 121846787-121846787 on Build GRCh38
rs772253268 Chr.12: 121846863-121846863 on Build GRCh38
rs189210271 Chr.12: 121846891-121846891 on Build GRCh38
rs372058146 Chr.12: 121846862-121846862 on Build GRCh38
rs756810403 Chr.12: 121846909-121846909 on Build GRCh38
rs545952069 Chr.12: 121846765-121846765 on Build GRCh38
rs967447098 Chr.12: 121846739-121846739 on Build GRCh38
rs759434513 Chr.12: 121846892-121846892 on Build GRCh38
rs779933779 Chr.12: 121846839-121846839 on Build GRCh38
rs998641456 Chr.12: 121846751-121846751 on Build GRCh38
rs369152577 Chr.12: 121846910-121846910 on Build GRCh38
rs749365084 Chr.12: 121846827-121846828 on Build GRCh38
rs755816788 Chr.12: 121846915-121846915 on Build GRCh38
rs375083142 Chr.12: 121846878-121846878 on Build GRCh38
rs373627459 Chr.12: 121846869-121846869 on Build GRCh38
rs144416002 Chr.12: 121846880-121846880 on Build GRCh38
rs780700651 Chr.12: 121846811-121846811 on Build GRCh38
rs369428110 Chr.12: 121846822-121846822 on Build GRCh38

View Full Product Details