Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29308570-29308811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2L1
Entrez Gene ID
10634
Gene Name
growth arrest specific 2 like 1
Gene Aliases
GAR22
UniGene
Hs.322852
Gene Chromosome Location
Chr.22: 29306618-29312789 on Build GRCh38
Transcripts
NM_001278730.1 NM_006478.4 NM_152236.2 XM_011529824.1 XM_011529825.1 XM_017028533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669382 c.561C>T p.T187T Chr.22: 29308666-29308666 on Build GRCh38
6196077 c.472G>A p.V158M Chr.22: 29308577-29308577 on Build GRCh38
3769880 c.511G>T p.A171S Chr.22: 29308616-29308616 on Build GRCh38
5859125 c.473T>C p.V158A Chr.22: 29308578-29308578 on Build GRCh38
3759107 c.532C>T p.P178S Chr.22: 29308637-29308637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772878158 Chr.22: 29308764-29308764 on Build GRCh38
rs950717626 Chr.22: 29308606-29308606 on Build GRCh38
rs755932088 Chr.22: 29308594-29308594 on Build GRCh38
rs746365930 Chr.22: 29308715-29308715 on Build GRCh38
rs368533422 Chr.22: 29308685-29308685 on Build GRCh38
rs191844093 Chr.22: 29308637-29308637 on Build GRCh38
rs148265669 Chr.22: 29308607-29308607 on Build GRCh38
rs567152329 Chr.22: 29308722-29308722 on Build GRCh38
rs777070349 Chr.22: 29308640-29308640 on Build GRCh38
rs770295311 Chr.22: 29308717-29308717 on Build GRCh38
rs541058137 Chr.22: 29308698-29308698 on Build GRCh38
rs778008336 Chr.22: 29308697-29308697 on Build GRCh38
rs769281251 Chr.22: 29308634-29308634 on Build GRCh38
rs930946576 Chr.22: 29308601-29308601 on Build GRCh38
rs762725485 Chr.22: 29308658-29308658 on Build GRCh38
rs753741281 Chr.22: 29308782-29308782 on Build GRCh38
rs750388383 Chr.22: 29308673-29308673 on Build GRCh38
rs372396375 Chr.22: 29308710-29308710 on Build GRCh38
rs766096399 Chr.22: 29308688-29308688 on Build GRCh38
rs1019814796 Chr.22: 29308768-29308768 on Build GRCh38
rs529843808 Chr.22: 29308771-29308771 on Build GRCh38
rs749720814 Chr.22: 29308729-29308729 on Build GRCh38
rs774216974 Chr.22: 29308755-29308755 on Build GRCh38
rs774125414 Chr.22: 29308667-29308667 on Build GRCh38
rs147637354 Chr.22: 29308770-29308770 on Build GRCh38
rs201596421 Chr.22: 29308652-29308652 on Build GRCh38
rs781274105 Chr.22: 29308702-29308702 on Build GRCh38
rs763791690 Chr.22: 29308666-29308666 on Build GRCh38
rs758574943 Chr.22: 29308619-29308619 on Build GRCh38
rs914189363 Chr.22: 29308713-29308713 on Build GRCh38
rs754742383 Chr.22: 29308696-29308696 on Build GRCh38
rs56037813 Chr.22: 29308672-29308672 on Build GRCh38
rs770067614 Chr.22: 29308647-29308647 on Build GRCh38
rs113516975 Chr.22: 29308609-29308609 on Build GRCh38
rs545449101 Chr.22: 29308591-29308591 on Build GRCh38
rs756025463 Chr.22: 29308683-29308683 on Build GRCh38
rs780305998 Chr.22: 29308721-29308721 on Build GRCh38
rs771498880 Chr.22: 29308766-29308766 on Build GRCh38
rs371060454 Chr.22: 29308708-29308708 on Build GRCh38
rs369870873 Chr.22: 29308628-29308628 on Build GRCh38
rs1011521148 Chr.22: 29308679-29308679 on Build GRCh38
rs1027634800 Chr.22: 29308785-29308785 on Build GRCh38
rs763124341 Chr.22: 29308760-29308779 on Build GRCh38
rs753617015 Chr.22: 29308692-29308692 on Build GRCh38
rs772715078 Chr.22: 29308769-29308769 on Build GRCh38
rs769113573 Chr.22: 29308743-29308743 on Build GRCh38
rs757383475 Chr.22: 29308701-29308701 on Build GRCh38
rs746371311 Chr.22: 29308642-29308642 on Build GRCh38
rs188482420 Chr.22: 29308633-29308633 on Build GRCh38

View Full Product Details