Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73483756-73484001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFM
Entrez Gene ID
173
Gene Name
afamin
Gene Aliases
ALB2, ALBA, ALF
UniGene
Hs.168718
Gene Chromosome Location
Chr.4: 73481740-73504001 on Build GRCh38
Transcripts
NM_001133.2 XM_017007842.1 XM_017007843.1 XM_017007844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1057426 c.90G>T p.E30D Chr.4: 73483942-73483942 on Build GRCh38
259348 c.110A>C p.K37T Chr.4: 73483962-73483962 on Build GRCh38
6100987 c.128T>C p.I43T Chr.4: 73483980-73483980 on Build GRCh38
6836345 c.89-1G>C p.? Chr.4: 73483940-73483940 on Build GRCh38
2831198 c.113T>G p.F38C Chr.4: 73483965-73483965 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs367555825 Chr.4: 73483924-73483924 on Build GRCh38
rs986298452 Chr.4: 73483883-73483883 on Build GRCh38
rs762960521 Chr.4: 73483949-73483949 on Build GRCh38
rs757636540 Chr.4: 73483954-73483954 on Build GRCh38
rs752012336 Chr.4: 73483951-73483951 on Build GRCh38
rs774177553 Chr.4: 73483910-73483910 on Build GRCh38
rs200353894 Chr.4: 73483950-73483950 on Build GRCh38
rs967906821 Chr.4: 73483888-73483899 on Build GRCh38
rs910479964 Chr.4: 73483839-73483839 on Build GRCh38
rs749677929 Chr.4: 73483919-73483919 on Build GRCh38
rs771862126 Chr.4: 73483920-73483920 on Build GRCh38
rs201597484 Chr.4: 73483939-73483939 on Build GRCh38
rs759815425 Chr.4: 73483921-73483921 on Build GRCh38
rs955002089 Chr.4: 73483857-73483857 on Build GRCh38
rs536487748 Chr.4: 73483830-73483830 on Build GRCh38
rs752731440 Chr.4: 73483931-73483931 on Build GRCh38
rs935700449 Chr.4: 73483801-73483801 on Build GRCh38
rs570053222 Chr.4: 73483867-73483867 on Build GRCh38
rs768599597 Chr.4: 73483903-73483903 on Build GRCh38
rs370431081 Chr.4: 73483920-73483920 on Build GRCh38
rs181509212 Chr.4: 73483807-73483807 on Build GRCh38
rs1030505473 Chr.4: 73483829-73483829 on Build GRCh38
rs914199372 Chr.4: 73483948-73483948 on Build GRCh38
rs200873818 Chr.4: 73483915-73483915 on Build GRCh38
rs781727350 Chr.4: 73483958-73483958 on Build GRCh38
rs750076443 Chr.4: 73483969-73483969 on Build GRCh38

View Full Product Details