Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203015915-203016188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM183A
Entrez Gene ID
92703
Gene Name
transmembrane protein 183A
Gene Aliases
C1orf37
UniGene
Hs.497443
Gene Chromosome Location
Chr.1: 203007367-203024848 on Build GRCh38
Transcripts
NM_001322955.1 NM_001322956.1 NM_001322957.1 NM_001322958.1 NM_001322959.1 NM_138391.4 NR_136530.1 NR_136531.1 NR_136532.1 NR_136533.1 NR_136534.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4576778 c.542A>G p.D181G Chr.1: 203015974-203015974 on Build GRCh38
300817 c.677A>G p.E226G Chr.1: 203016109-203016109 on Build GRCh38
6803608 c.536C>T p.T179M Chr.1: 203015968-203015968 on Build GRCh38
6803609 c.649C>T p.R217* Chr.1: 203016081-203016081 on Build GRCh38
4824187 c.656C>T p.S219F Chr.1: 203016088-203016088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773123898 Chr.1: 203016126-203016126 on Build GRCh38
rs760779263 Chr.1: 203015962-203015962 on Build GRCh38
rs776708032 Chr.1: 203016002-203016003 on Build GRCh38
rs546957992 Chr.1: 203016021-203016021 on Build GRCh38
rs374714789 Chr.1: 203016147-203016147 on Build GRCh38
rs749124727 Chr.1: 203016124-203016124 on Build GRCh38
rs775416962 Chr.1: 203016120-203016120 on Build GRCh38
rs776372506 Chr.1: 203015990-203015990 on Build GRCh38
rs745560644 Chr.1: 203016111-203016111 on Build GRCh38
rs759000597 Chr.1: 203016088-203016088 on Build GRCh38
rs754257024 Chr.1: 203015943-203015943 on Build GRCh38
rs780801756 Chr.1: 203016107-203016107 on Build GRCh38
rs893773840 Chr.1: 203016031-203016031 on Build GRCh38
rs148817359 Chr.1: 203016008-203016008 on Build GRCh38
rs934511644 Chr.1: 203015952-203015952 on Build GRCh38
rs746561575 Chr.1: 203016096-203016096 on Build GRCh38
rs946700813 Chr.1: 203016095-203016095 on Build GRCh38
rs777407353 Chr.1: 203015969-203015969 on Build GRCh38
rs769577143 Chr.1: 203016113-203016113 on Build GRCh38
rs777087124 Chr.1: 203016091-203016091 on Build GRCh38
rs567022563 Chr.1: 203016125-203016125 on Build GRCh38
rs773588361 Chr.1: 203015950-203015950 on Build GRCh38
rs755500011 Chr.1: 203015945-203015945 on Build GRCh38
rs748734117 Chr.1: 203015960-203015960 on Build GRCh38
rs371737477 Chr.1: 203016101-203016101 on Build GRCh38
rs765737467 Chr.1: 203016058-203016058 on Build GRCh38
rs367857593 Chr.1: 203015968-203015968 on Build GRCh38
rs760744206 Chr.1: 203016016-203016016 on Build GRCh38
rs775375578 Chr.1: 203015998-203015998 on Build GRCh38
rs754203775 Chr.1: 203016025-203016025 on Build GRCh38
rs779581280 Chr.1: 203015953-203015953 on Build GRCh38
rs193920903 Chr.1: 203016141-203016141 on Build GRCh38
rs760828714 Chr.1: 203016146-203016146 on Build GRCh38
rs746627540 Chr.1: 203015981-203015981 on Build GRCh38
rs755367985 Chr.1: 203016030-203016030 on Build GRCh38
rs770582206 Chr.1: 203015987-203015987 on Build GRCh38
rs753928593 Chr.1: 203016005-203016005 on Build GRCh38
rs527303210 Chr.1: 203016009-203016009 on Build GRCh38
rs753139412 Chr.1: 203016071-203016071 on Build GRCh38
rs769626435 Chr.1: 203015991-203015991 on Build GRCh38
rs935380024 Chr.1: 203016018-203016018 on Build GRCh38
rs776958564 Chr.1: 203016157-203016157 on Build GRCh38
rs766483812 Chr.1: 203015974-203015974 on Build GRCh38

View Full Product Details