Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 20164700-20164959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC18A1
Entrez Gene ID
6570
Gene Name
solute carrier family 18 member A1
Gene Aliases
CGAT, VAT1, VMAT1
UniGene
Hs.158322
Gene Chromosome Location
Chr.8: 20144855-20183206 on Build GRCh38
Transcripts
NM_001135691.2 NM_001142324.1 NM_001142325.1 NM_003053.3 XM_011544625.1 XM_011544626.1 XM_011544627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1699780 c.978G>A p.W326* Chr.8: 20164906-20164906 on Build GRCh38
6005990 c.942G>T p.M314I Chr.8: 20164942-20164942 on Build GRCh38
3924981 c.989C>T p.T330I Chr.8: 20164895-20164895 on Build GRCh38
2150029 c.931T>A p.F311I Chr.8: 20164953-20164953 on Build GRCh38
4788850 c.925A>G p.I309V Chr.8: 20164959-20164959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043161150 Chr.8: 20164744-20164744 on Build GRCh38
rs1041746084 Chr.8: 20164852-20164852 on Build GRCh38
rs758734675 Chr.8: 20164825-20164827 on Build GRCh38
rs781604557 Chr.8: 20164837-20164837 on Build GRCh38
rs763824942 Chr.8: 20164871-20164871 on Build GRCh38
rs202033958 Chr.8: 20164884-20164884 on Build GRCh38
rs773563931 Chr.8: 20164848-20164848 on Build GRCh38
rs768462280 Chr.8: 20164927-20164927 on Build GRCh38
rs778173619 Chr.8: 20164937-20164938 on Build GRCh38
rs747536359 Chr.8: 20164834-20164834 on Build GRCh38
rs960328633 Chr.8: 20164849-20164849 on Build GRCh38
rs375980427 Chr.8: 20164917-20164917 on Build GRCh38
rs371723932 Chr.8: 20164842-20164842 on Build GRCh38
rs150790381 Chr.8: 20164886-20164886 on Build GRCh38
rs937530938 Chr.8: 20164818-20164818 on Build GRCh38
rs760239736 Chr.8: 20164926-20164926 on Build GRCh38
rs866366104 Chr.8: 20164756-20164756 on Build GRCh38
rs771556783 Chr.8: 20164919-20164919 on Build GRCh38
rs763465902 Chr.8: 20164851-20164851 on Build GRCh38
rs1053646585 Chr.8: 20164893-20164893 on Build GRCh38
rs534807467 Chr.8: 20164737-20164737 on Build GRCh38
rs761480509 Chr.8: 20164882-20164882 on Build GRCh38
rs1036203159 Chr.8: 20164780-20164780 on Build GRCh38
rs1029181374 Chr.8: 20164753-20164753 on Build GRCh38
rs200134564 Chr.8: 20164847-20164847 on Build GRCh38
rs776177783 Chr.8: 20164929-20164929 on Build GRCh38
rs760466707 Chr.8: 20164869-20164869 on Build GRCh38
rs761479990 Chr.8: 20164938-20164938 on Build GRCh38
rs757835343 Chr.8: 20164804-20164804 on Build GRCh38
rs781373716 Chr.8: 20164904-20164904 on Build GRCh38
rs144890324 Chr.8: 20164754-20164754 on Build GRCh38
rs13248157 Chr.8: 20164799-20164799 on Build GRCh38
rs780112295 Chr.8: 20164892-20164892 on Build GRCh38
rs141032586 Chr.8: 20164742-20164742 on Build GRCh38
rs1028817950 Chr.8: 20164918-20164918 on Build GRCh38
rs775198550 Chr.8: 20164923-20164923 on Build GRCh38
rs1009105015 Chr.8: 20164774-20164774 on Build GRCh38
rs769109299 Chr.8: 20164872-20164872 on Build GRCh38
rs770088407 Chr.8: 20164913-20164913 on Build GRCh38
rs969433518 Chr.8: 20164814-20164814 on Build GRCh38
rs751635329 Chr.8: 20164896-20164896 on Build GRCh38
rs771720348 Chr.8: 20164864-20164864 on Build GRCh38
rs755254673 Chr.8: 20164902-20164902 on Build GRCh38
rs764957602 Chr.8: 20164939-20164939 on Build GRCh38
rs539178176 Chr.8: 20164782-20164782 on Build GRCh38
rs995997699 Chr.8: 20164750-20164750 on Build GRCh38
rs200896623 Chr.8: 20164911-20164911 on Build GRCh38
rs748589025 Chr.8: 20164906-20164906 on Build GRCh38
rs79782093 Chr.8: 20164805-20164805 on Build GRCh38
rs920824365 Chr.8: 20164806-20164806 on Build GRCh38
rs768803744 Chr.8: 20164832-20164832 on Build GRCh38

View Full Product Details