Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45469723-45469970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WFDC2
Entrez Gene ID
10406
Gene Name
WAP four-disulfide core domain 2
Gene Aliases
EDDM4, HE4, WAP5, dJ461P17.6
UniGene
Hs.2719
Gene Chromosome Location
Chr.20: 45469754-45481532 on Build GRCh38
Transcripts
NM_006103.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4648402 c.23C>T p.P8L Chr.20: 45469804-45469804 on Build GRCh38
4741160 c.3G>A p.M1I Chr.20: 45469784-45469784 on Build GRCh38
1027256 c.7G>A p.A3T Chr.20: 45469788-45469788 on Build GRCh38
3547031 c.40C>T p.L14F Chr.20: 45469821-45469821 on Build GRCh38
4627816 c.14G>A p.R5H Chr.20: 45469795-45469795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750036554 Chr.20: 45469899-45469899 on Build GRCh38
rs773479617 Chr.20: 45469851-45469851 on Build GRCh38
rs773424760 Chr.20: 45469784-45469784 on Build GRCh38
rs768351013 Chr.20: 45469826-45469826 on Build GRCh38
rs567986690 Chr.20: 45469891-45469891 on Build GRCh38
rs981539501 Chr.20: 45469919-45469919 on Build GRCh38
rs909489389 Chr.20: 45469925-45469925 on Build GRCh38
rs751456354 Chr.20: 45469910-45469910 on Build GRCh38
rs775815158 Chr.20: 45469887-45469887 on Build GRCh38
rs564004546 Chr.20: 45469906-45469906 on Build GRCh38
rs11545050 Chr.20: 45469816-45469816 on Build GRCh38
rs981507624 Chr.20: 45469904-45469904 on Build GRCh38
rs748449364 Chr.20: 45469842-45469842 on Build GRCh38
rs111895648 Chr.20: 45469862-45469862 on Build GRCh38
rs750981141 Chr.20: 45469743-45469743 on Build GRCh38
rs573015719 Chr.20: 45469923-45469923 on Build GRCh38
rs763487678 Chr.20: 45469747-45469747 on Build GRCh38
rs749912153 Chr.20: 45469817-45469817 on Build GRCh38
rs746748464 Chr.20: 45469777-45469777 on Build GRCh38
rs774129788 Chr.20: 45469817-45469819 on Build GRCh38
rs745869262 Chr.20: 45469774-45469774 on Build GRCh38
rs759837646 Chr.20: 45469796-45469796 on Build GRCh38
rs1020106238 Chr.20: 45469894-45469894 on Build GRCh38
rs764838282 Chr.20: 45469893-45469893 on Build GRCh38
rs763617796 Chr.20: 45469785-45469785 on Build GRCh38
rs6017577 Chr.20: 45469849-45469849 on Build GRCh38
rs766777303 Chr.20: 45469793-45469793 on Build GRCh38
rs758370821 Chr.20: 45469820-45469820 on Build GRCh38
rs75766050 Chr.20: 45469902-45469902 on Build GRCh38
rs372965919 Chr.20: 45469783-45469783 on Build GRCh38
rs767862116 Chr.20: 45469798-45469798 on Build GRCh38
rs779885365 Chr.20: 45469821-45469821 on Build GRCh38
rs200690352 Chr.20: 45469762-45469762 on Build GRCh38
rs749350035 Chr.20: 45469859-45469859 on Build GRCh38
rs540030806 Chr.20: 45469952-45469952 on Build GRCh38
rs766922421 Chr.20: 45469748-45469748 on Build GRCh38
rs761687086 Chr.20: 45469878-45469889 on Build GRCh38
rs760018642 Chr.20: 45469874-45469874 on Build GRCh38
rs751997990 Chr.20: 45469794-45469794 on Build GRCh38
rs774730344 Chr.20: 45469867-45469867 on Build GRCh38
rs537635906 Chr.20: 45469833-45469833 on Build GRCh38
rs200042356 Chr.20: 45469775-45469775 on Build GRCh38
rs771006413 Chr.20: 45469866-45469866 on Build GRCh38
rs756895288 Chr.20: 45469814-45469814 on Build GRCh38
rs746917053 Chr.20: 45469823-45469823 on Build GRCh38
rs942285551 Chr.20: 45469933-45469933 on Build GRCh38
rs780818562 Chr.20: 45469752-45469752 on Build GRCh38
rs773110714 Chr.20: 45469897-45469908 on Build GRCh38
rs765858989 Chr.20: 45469907-45469907 on Build GRCh38
rs370328706 Chr.20: 45469763-45469763 on Build GRCh38
rs756770584 Chr.20: 45469765-45469765 on Build GRCh38
rs897991657 Chr.20: 45469945-45469945 on Build GRCh38
rs776743061 Chr.20: 45469782-45469782 on Build GRCh38
rs753433683 Chr.20: 45469804-45469804 on Build GRCh38
rs957265455 Chr.20: 45469883-45469883 on Build GRCh38
rs767831930 Chr.20: 45469875-45469875 on Build GRCh38
rs778323797 Chr.20: 45469772-45469772 on Build GRCh38
rs201928970 Chr.20: 45469832-45469832 on Build GRCh38
rs1036973348 Chr.20: 45469936-45469936 on Build GRCh38
rs879122786 Chr.20: 45469861-45469862 on Build GRCh38
rs1006774733 Chr.20: 45469827-45469827 on Build GRCh38
rs552243960 Chr.20: 45469781-45469781 on Build GRCh38
rs6032215 Chr.20: 45469757-45469757 on Build GRCh38
rs749331629 Chr.20: 45469766-45469766 on Build GRCh38
rs771945939 Chr.20: 45469891-45469893 on Build GRCh38
rs374994072 Chr.20: 45469914-45469914 on Build GRCh38
rs751860098 Chr.20: 45469761-45469761 on Build GRCh38
rs879045598 Chr.20: 45469863-45469863 on Build GRCh38

View Full Product Details