Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39514247-39514478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIEF1
Entrez Gene ID
54471
Gene Name
mitochondrial elongation factor 1
Gene Aliases
HSU79252, MID51, SMCR7L, dJ1104E15.3
UniGene
Hs.728085
Gene Chromosome Location
Chr.22: 39500100-39518134 on Build GRCh38
Transcripts
NM_001304564.1 NM_019008.5 NR_130789.1 XM_011530236.1 XM_011530237.1 XM_011530238.1 XM_017028838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4104247 c.1389G>A p.T463T Chr.22: 39514320-39514320 on Build GRCh38
1034308 c.1324G>T p.E442* Chr.22: 39514255-39514255 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs142355530 Chr.22: 39514320-39514320 on Build GRCh38
rs747332598 Chr.22: 39514342-39514342 on Build GRCh38
rs774289422 Chr.22: 39514341-39514341 on Build GRCh38
rs767615419 Chr.22: 39514405-39514405 on Build GRCh38
rs753766036 Chr.22: 39514302-39514302 on Build GRCh38
rs750676080 Chr.22: 39514324-39514324 on Build GRCh38
rs974948306 Chr.22: 39514410-39514410 on Build GRCh38
rs922014236 Chr.22: 39514391-39514391 on Build GRCh38
rs767234558 Chr.22: 39514281-39514281 on Build GRCh38
rs760530375 Chr.22: 39514295-39514295 on Build GRCh38
rs752475555 Chr.22: 39514289-39514289 on Build GRCh38
rs373709239 Chr.22: 39514303-39514303 on Build GRCh38
rs973948002 Chr.22: 39514379-39514380 on Build GRCh38
rs187514447 Chr.22: 39514315-39514315 on Build GRCh38
rs369984275 Chr.22: 39514373-39514373 on Build GRCh38
rs912033199 Chr.22: 39514392-39514392 on Build GRCh38
rs369207595 Chr.22: 39514299-39514299 on Build GRCh38
rs553210294 Chr.22: 39514344-39514344 on Build GRCh38
rs770594362 Chr.22: 39514360-39514360 on Build GRCh38
rs748633276 Chr.22: 39514347-39514347 on Build GRCh38
rs779859266 Chr.22: 39514375-39514378 on Build GRCh38
rs943653683 Chr.22: 39514395-39514395 on Build GRCh38
rs2232095 Chr.22: 39514372-39514372 on Build GRCh38
rs376185243 Chr.22: 39514338-39514338 on Build GRCh38
rs935305275 Chr.22: 39514445-39514445 on Build GRCh38
rs768888462 Chr.22: 39514343-39514343 on Build GRCh38
rs758600363 Chr.22: 39514326-39514326 on Build GRCh38
rs759121747 Chr.22: 39514366-39514366 on Build GRCh38
rs762638970 Chr.22: 39514276-39514276 on Build GRCh38
rs149438399 Chr.22: 39514319-39514319 on Build GRCh38
rs925986226 Chr.22: 39514422-39514422 on Build GRCh38
rs773946058 Chr.22: 39514364-39514364 on Build GRCh38

View Full Product Details