Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84193803-84194070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAD2
Entrez Gene ID
161931
Gene Name
adenosine deaminase domain containing 2
Gene Aliases
TENRL
UniGene
Hs.8977
Gene Chromosome Location
Chr.16: 84191117-84197166 on Build GRCh38
Transcripts
NM_001145400.1 NM_139174.3 XM_005255814.1 XM_011522919.1 XM_011522920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4758285 c.471T>G p.C157W Chr.16: 84194058-84194058 on Build GRCh38
3512941 c.454G>A p.E152K Chr.16: 84194041-84194041 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751627781 Chr.16: 84194003-84194003 on Build GRCh38
rs563298953 Chr.16: 84193834-84193834 on Build GRCh38
rs747683078 Chr.16: 84194029-84194029 on Build GRCh38
rs1011224702 Chr.16: 84193955-84193955 on Build GRCh38
rs374748517 Chr.16: 84194024-84194024 on Build GRCh38
rs566713534 Chr.16: 84193961-84193961 on Build GRCh38
rs772855939 Chr.16: 84194038-84194038 on Build GRCh38
rs191653737 Chr.16: 84193844-84193844 on Build GRCh38
rs930190556 Chr.16: 84193860-84193860 on Build GRCh38
rs767178739 Chr.16: 84193979-84193979 on Build GRCh38
rs968496845 Chr.16: 84193908-84193908 on Build GRCh38
rs906577263 Chr.16: 84193904-84193904 on Build GRCh38
rs559191629 Chr.16: 84193916-84193916 on Build GRCh38
rs147117105 Chr.16: 84193918-84193918 on Build GRCh38
rs745714480 Chr.16: 84194011-84194011 on Build GRCh38
rs201533129 Chr.16: 84193975-84193975 on Build GRCh38
rs956723430 Chr.16: 84193858-84193858 on Build GRCh38
rs531140020 Chr.16: 84194022-84194022 on Build GRCh38
rs1012459366 Chr.16: 84193889-84193889 on Build GRCh38
rs1003649027 Chr.16: 84193906-84193906 on Build GRCh38
rs769467816 Chr.16: 84193967-84193968 on Build GRCh38
rs759833577 Chr.16: 84194044-84194044 on Build GRCh38
rs879241428 Chr.16: 84194051-84194051 on Build GRCh38
rs759942759 Chr.16: 84193995-84193995 on Build GRCh38
rs545958626 Chr.16: 84193890-84193890 on Build GRCh38
rs985891240 Chr.16: 84193964-84193964 on Build GRCh38
rs773904421 Chr.16: 84194028-84194028 on Build GRCh38
rs568626717 Chr.16: 84194010-84194010 on Build GRCh38
rs769509002 Chr.16: 84194012-84194012 on Build GRCh38
rs942537329 Chr.16: 84193981-84193981 on Build GRCh38
rs953208782 Chr.16: 84193963-84193963 on Build GRCh38
rs762450816 Chr.16: 84193959-84193959 on Build GRCh38
rs1022209841 Chr.16: 84193957-84193957 on Build GRCh38
rs888596375 Chr.16: 84193870-84193870 on Build GRCh38
rs1023818551 Chr.16: 84193895-84193895 on Build GRCh38
rs771692615 Chr.16: 84194030-84194030 on Build GRCh38
rs781428906 Chr.16: 84193983-84193983 on Build GRCh38
rs772819566 Chr.16: 84194031-84194031 on Build GRCh38
rs956762987 Chr.16: 84193951-84193951 on Build GRCh38
rs548516997 Chr.16: 84193931-84193931 on Build GRCh38
rs753198184 Chr.16: 84194001-84194001 on Build GRCh38
rs368019635 Chr.16: 84194026-84194026 on Build GRCh38
rs200616300 Chr.16: 84193971-84193971 on Build GRCh38
rs1048669448 Chr.16: 84193869-84193869 on Build GRCh38
rs371302161 Chr.16: 84193977-84193977 on Build GRCh38
rs758575211 Chr.16: 84194002-84194002 on Build GRCh38
rs915331954 Chr.16: 84193898-84193898 on Build GRCh38
rs754217473 Chr.16: 84193989-84193989 on Build GRCh38
rs774026785 Chr.16: 84193973-84193973 on Build GRCh38

View Full Product Details