Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5227209-5227442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WIPI2
Entrez Gene ID
26100
Gene Name
WD repeat domain, phosphoinositide interacting 2
Gene Aliases
ATG18B, Atg21, CGI-50, WIPI-2
UniGene
Hs.122363
Gene Chromosome Location
Chr.7: 5190204-5233855 on Build GRCh38
Transcripts
NM_001033518.1 NM_001033519.1 NM_001033520.1 NM_001278299.1 NM_015610.3 NM_016003.3 XM_006715685.1 XM_017011952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6822760 c.709G>A p.G237R Chr.7: 5227217-5227217 on Build GRCh38
6822775 c.960C>T p.F320F Chr.7: 5227291-5227291 on Build GRCh38
5165903 c.708C>T p.F236F Chr.7: 5227216-5227216 on Build GRCh38
6822764 c.789G>A p.T263T Chr.7: 5227297-5227297 on Build GRCh38
3881707 c.943A>G p.N315D Chr.7: 5227274-5227274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556202972 Chr.7: 5227409-5227409 on Build GRCh38
rs754666928 Chr.7: 5227391-5227391 on Build GRCh38
rs566274344 Chr.7: 5227270-5227270 on Build GRCh38
rs368759450 Chr.7: 5227377-5227377 on Build GRCh38
rs373112670 Chr.7: 5227291-5227291 on Build GRCh38
rs749896284 Chr.7: 5227378-5227378 on Build GRCh38
rs764581414 Chr.7: 5227368-5227368 on Build GRCh38
rs138529175 Chr.7: 5227275-5227275 on Build GRCh38
rs942458247 Chr.7: 5227415-5227415 on Build GRCh38
rs764279464 Chr.7: 5227302-5227302 on Build GRCh38
rs200673321 Chr.7: 5227366-5227366 on Build GRCh38
rs371146538 Chr.7: 5227315-5227315 on Build GRCh38
rs1134491 Chr.7: 5227288-5227288 on Build GRCh38
rs78789321 Chr.7: 5227302-5227302 on Build GRCh38
rs201087277 Chr.7: 5227357-5227357 on Build GRCh38
rs967438840 Chr.7: 5227400-5227400 on Build GRCh38
rs995731231 Chr.7: 5227346-5227346 on Build GRCh38
rs751991726 Chr.7: 5227369-5227369 on Build GRCh38
rs763418262 Chr.7: 5227299-5227299 on Build GRCh38
rs373378911 Chr.7: 5227360-5227360 on Build GRCh38
rs778658697 Chr.7: 5227392-5227392 on Build GRCh38
rs28510502 Chr.7: 5227359-5227359 on Build GRCh38
rs766146632 Chr.7: 5227339-5227339 on Build GRCh38
rs751610174 Chr.7: 5227368-5227371 on Build GRCh38
rs368546108 Chr.7: 5227322-5227322 on Build GRCh38
rs754839171 Chr.7: 5227412-5227412 on Build GRCh38
rs375023285 Chr.7: 5227344-5227344 on Build GRCh38
rs1047299264 Chr.7: 5227356-5227356 on Build GRCh38
rs768451585 Chr.7: 5227336-5227336 on Build GRCh38
rs757714583 Chr.7: 5227403-5227403 on Build GRCh38
rs773023213 Chr.7: 5227312-5227312 on Build GRCh38
rs776560982 Chr.7: 5227353-5227353 on Build GRCh38
rs751565385 Chr.7: 5227410-5227410 on Build GRCh38
rs749014711 Chr.7: 5227330-5227330 on Build GRCh38
rs890278181 Chr.7: 5227399-5227399 on Build GRCh38
rs982607564 Chr.7: 5227323-5227323 on Build GRCh38
rs916597059 Chr.7: 5227279-5227279 on Build GRCh38
rs762200166 Chr.7: 5227371-5227371 on Build GRCh38
rs1030847519 Chr.7: 5227413-5227413 on Build GRCh38
rs150877444 Chr.7: 5227297-5227297 on Build GRCh38
rs753877120 Chr.7: 5227259-5227259 on Build GRCh38
rs753509034 Chr.7: 5227386-5227386 on Build GRCh38
rs770949051 Chr.7: 5227252-5227252 on Build GRCh38
rs372852276 Chr.7: 5227384-5227384 on Build GRCh38
rs772096205 Chr.7: 5227351-5227351 on Build GRCh38
rs748005611 Chr.7: 5227345-5227345 on Build GRCh38
rs755625785 Chr.7: 5227383-5227383 on Build GRCh38
rs28605777 Chr.7: 5227327-5227327 on Build GRCh38
rs780533277 Chr.7: 5227316-5227316 on Build GRCh38
rs765617109 Chr.7: 5227295-5227295 on Build GRCh38

View Full Product Details