Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87529445-87529718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RARS2
Entrez Gene ID
57038
Gene Name
arginyl-tRNA synthetase 2, mitochondrial
Gene Aliases
ArgRS, DALRD2, PCH6, PRO1992, RARSL
UniGene
Hs.485910
Gene Chromosome Location
Chr.6: 87513744-87590032 on Build GRCh38
Transcripts
NM_001318785.1 NM_020320.4 NR_134857.1 XM_011535948.2 XM_011535949.2 XM_017011072.1 XM_017011073.1 XM_017011074.1 XM_017011075.1 XM_017011076.1 XM_017011077.1 XM_017011078.1 XM_017011079.1 XM_017011080.1 XM_017011081.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6763012 c.817C>T p.R273C Chr.6: 87529603-87529603 on Build GRCh38
3430934 c.808T>A p.S270T Chr.6: 87529612-87529612 on Build GRCh38
7218634 c.872A>G p.K291R Chr.6: 87529548-87529548 on Build GRCh38
3394342 c.878+5G>T p.? Chr.6: 87529537-87529537 on Build GRCh38
5346546 c.800C>T p.S267L Chr.6: 87529620-87529620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757746207 Chr.6: 87529508-87529508 on Build GRCh38
rs369331184 Chr.6: 87529556-87529556 on Build GRCh38
rs774075596 Chr.6: 87529638-87529638 on Build GRCh38
rs550849175 Chr.6: 87529574-87529574 on Build GRCh38
rs763781048 Chr.6: 87529528-87529528 on Build GRCh38
rs758720333 Chr.6: 87529515-87529515 on Build GRCh38
rs1002729156 Chr.6: 87529642-87529642 on Build GRCh38
rs188700495 Chr.6: 87529523-87529523 on Build GRCh38
rs774044535 Chr.6: 87529535-87529535 on Build GRCh38
rs201214613 Chr.6: 87529525-87529525 on Build GRCh38
rs756586121 Chr.6: 87529499-87529499 on Build GRCh38
rs770878816 Chr.6: 87529637-87529637 on Build GRCh38
rs758622612 Chr.6: 87529591-87529591 on Build GRCh38
rs139721632 Chr.6: 87529602-87529602 on Build GRCh38
rs777737159 Chr.6: 87529603-87529603 on Build GRCh38
rs1013045059 Chr.6: 87529570-87529570 on Build GRCh38
rs1036631557 Chr.6: 87529496-87529496 on Build GRCh38
rs541352251 Chr.6: 87529587-87529587 on Build GRCh38
rs919642361 Chr.6: 87529660-87529660 on Build GRCh38
rs863224186 Chr.6: 87529648-87529648 on Build GRCh38
rs764639091 Chr.6: 87529568-87529568 on Build GRCh38
rs749185497 Chr.6: 87529630-87529630 on Build GRCh38
rs780553688 Chr.6: 87529651-87529651 on Build GRCh38
rs771742364 Chr.6: 87529651-87529651 on Build GRCh38
rs774161360 Chr.6: 87529542-87529542 on Build GRCh38
rs372378738 Chr.6: 87529509-87529509 on Build GRCh38
rs774997689 Chr.6: 87529551-87529551 on Build GRCh38
rs113367519 Chr.6: 87529565-87529565 on Build GRCh38
rs367622665 Chr.6: 87529689-87529689 on Build GRCh38
rs17850652 Chr.6: 87529548-87529548 on Build GRCh38
rs886670267 Chr.6: 87529471-87529471 on Build GRCh38
rs757868591 Chr.6: 87529584-87529584 on Build GRCh38
rs368859792 Chr.6: 87529651-87529652 on Build GRCh38
rs759139003 Chr.6: 87529544-87529544 on Build GRCh38
rs775053238 Chr.6: 87529657-87529657 on Build GRCh38
rs764576122 Chr.6: 87529502-87529502 on Build GRCh38
rs751179046 Chr.6: 87529585-87529585 on Build GRCh38
rs145297855 Chr.6: 87529647-87529647 on Build GRCh38
rs770750240 Chr.6: 87529537-87529537 on Build GRCh38
rs746050674 Chr.6: 87529510-87529510 on Build GRCh38
rs146607200 Chr.6: 87529505-87529505 on Build GRCh38
rs780127098 Chr.6: 87529518-87529518 on Build GRCh38

View Full Product Details