Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24206302-24206567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSSK4
Entrez Gene ID
283629
Gene Name
testis specific serine kinase 4
Gene Aliases
C14orf20, STK22E, TSK-4, TSK4, TSSK5
UniGene
Hs.314432
Gene Chromosome Location
Chr.14: 24205524-24208362 on Build GRCh38
Transcripts
NM_001184739.1 NM_001308067.1 NM_174944.3 XM_011536663.2 XM_017021226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3495385 c.231G>A p.M77I Chr.14: 24206514-24206514 on Build GRCh38
4050213 c.234A>C p.K78N Chr.14: 24206517-24206517 on Build GRCh38
5847932 c.241C>T p.R81W Chr.14: 24206524-24206524 on Build GRCh38
4050214 c.234A>C p.K78N Chr.14: 24206517-24206517 on Build GRCh38
3401258 c.275T>C p.I92T Chr.14: 24206558-24206558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562899477 Chr.14: 24206469-24206469 on Build GRCh38
rs758953254 Chr.14: 24206482-24206482 on Build GRCh38
rs774910961 Chr.14: 24206377-24206377 on Build GRCh38
rs913772093 Chr.14: 24206364-24206364 on Build GRCh38
rs978757724 Chr.14: 24206325-24206325 on Build GRCh38
rs756803813 Chr.14: 24206471-24206471 on Build GRCh38
rs748379677 Chr.14: 24206505-24206505 on Build GRCh38
rs755852868 Chr.14: 24206397-24206397 on Build GRCh38
rs763099307 Chr.14: 24206538-24206538 on Build GRCh38
rs932320004 Chr.14: 24206432-24206432 on Build GRCh38
rs768944691 Chr.14: 24206503-24206503 on Build GRCh38
rs751030358 Chr.14: 24206460-24206460 on Build GRCh38
rs1032299215 Chr.14: 24206465-24206465 on Build GRCh38
rs768283998 Chr.14: 24206426-24206426 on Build GRCh38
rs371391834 Chr.14: 24206477-24206477 on Build GRCh38
rs1014384816 Chr.14: 24206495-24206495 on Build GRCh38
rs761637173 Chr.14: 24206486-24206486 on Build GRCh38
rs920843550 Chr.14: 24206410-24206410 on Build GRCh38
rs563942506 Chr.14: 24206385-24206385 on Build GRCh38
rs571823153 Chr.14: 24206339-24206339 on Build GRCh38
rs78386085 Chr.14: 24206401-24206401 on Build GRCh38
rs747148578 Chr.14: 24206362-24206362 on Build GRCh38
rs200401327 Chr.14: 24206524-24206524 on Build GRCh38
rs1003946871 Chr.14: 24206453-24206453 on Build GRCh38
rs1242017 Chr.14: 24206363-24206363 on Build GRCh38
rs780491244 Chr.14: 24206484-24206484 on Build GRCh38
rs938273272 Chr.14: 24206387-24206387 on Build GRCh38
rs776194487 Chr.14: 24206447-24206447 on Build GRCh38
rs781493676 Chr.14: 24206504-24206504 on Build GRCh38
rs780866608 Chr.14: 24206468-24206469 on Build GRCh38
rs367939541 Chr.14: 24206462-24206462 on Build GRCh38
rs763836721 Chr.14: 24206466-24206466 on Build GRCh38
rs931019831 Chr.14: 24206429-24206429 on Build GRCh38
rs754618259 Chr.14: 24206464-24206468 on Build GRCh38
rs769428073 Chr.14: 24206488-24206488 on Build GRCh38
rs745351988 Chr.14: 24206481-24206481 on Build GRCh38
rs1040859381 Chr.14: 24206449-24206449 on Build GRCh38
rs747407488 Chr.14: 24206490-24206490 on Build GRCh38
rs549848282 Chr.14: 24206446-24206446 on Build GRCh38
rs970471539 Chr.14: 24206324-24206324 on Build GRCh38
rs373553186 Chr.14: 24206337-24206337 on Build GRCh38

View Full Product Details