Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88428857-88429130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF469
Entrez Gene ID
84627
Gene Name
zinc finger protein 469
Gene Aliases
BCS, BCS1
UniGene
Hs.54925
Gene Chromosome Location
Chr.16: 88424805-88440757 on Build GRCh38
Transcripts
NM_001127464.2 XM_017023784.1 XM_017023785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
225912 c.1429C>T p.P477S Chr.16: 88428899-88428899 on Build GRCh38
5507056 c.1398C>T p.G466G Chr.16: 88428868-88428868 on Build GRCh38
6990258 c.1554A>T p.Q518H Chr.16: 88429024-88429024 on Build GRCh38
1234108 c.1453C>G p.P485A Chr.16: 88428923-88428923 on Build GRCh38
4528454 c.1538G>A p.W513* Chr.16: 88429008-88429008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759524656 Chr.16: 88428936-88428936 on Build GRCh38
rs776840891 Chr.16: 88428919-88428919 on Build GRCh38
rs939220494 Chr.16: 88428928-88428928 on Build GRCh38
rs953454298 Chr.16: 88429004-88429004 on Build GRCh38
rs202205643 Chr.16: 88428953-88428953 on Build GRCh38
rs1025580206 Chr.16: 88429033-88429033 on Build GRCh38
rs770926082 Chr.16: 88428876-88428876 on Build GRCh38
rs868468606 Chr.16: 88428944-88428944 on Build GRCh38
rs767072775 Chr.16: 88428977-88428977 on Build GRCh38
rs540133783 Chr.16: 88428914-88428914 on Build GRCh38
rs1003484667 Chr.16: 88429018-88429018 on Build GRCh38
rs117555121 Chr.16: 88428941-88428941 on Build GRCh38
rs184458982 Chr.16: 88429079-88429079 on Build GRCh38
rs969357442 Chr.16: 88428887-88428887 on Build GRCh38
rs758781186 Chr.16: 88429020-88429020 on Build GRCh38
rs143852982 Chr.16: 88429054-88429054 on Build GRCh38
rs189476639 Chr.16: 88429085-88429085 on Build GRCh38
rs1016285174 Chr.16: 88428929-88428929 on Build GRCh38
rs763933273 Chr.16: 88428958-88428958 on Build GRCh38
rs550263442 Chr.16: 88428978-88428978 on Build GRCh38
rs775578805 Chr.16: 88428888-88428888 on Build GRCh38
rs769822891 Chr.16: 88428937-88428937 on Build GRCh38
rs963422027 Chr.16: 88428934-88428934 on Build GRCh38
rs778051482 Chr.16: 88429022-88429022 on Build GRCh38
rs1052914598 Chr.16: 88429003-88429003 on Build GRCh38
rs28723506 Chr.16: 88428959-88428959 on Build GRCh38
rs281865145 Chr.16: 88429097-88429097 on Build GRCh38
rs755555951 Chr.16: 88428998-88428998 on Build GRCh38
rs1013774844 Chr.16: 88429026-88429026 on Build GRCh38
rs966608916 Chr.16: 88428996-88428996 on Build GRCh38
rs902329062 Chr.16: 88428906-88428906 on Build GRCh38
rs1042676401 Chr.16: 88428991-88428991 on Build GRCh38
rs750005577 Chr.16: 88428992-88428992 on Build GRCh38
rs200675426 Chr.16: 88428994-88428994 on Build GRCh38
rs753160761 Chr.16: 88429017-88429017 on Build GRCh38
rs7199961 Chr.16: 88428999-88428999 on Build GRCh38
rs761461530 Chr.16: 88428969-88428969 on Build GRCh38
rs919081661 Chr.16: 88428945-88428945 on Build GRCh38
rs978404383 Chr.16: 88429078-88429078 on Build GRCh38
rs865985857 Chr.16: 88429034-88429034 on Build GRCh38
rs1056103582 Chr.16: 88428933-88428933 on Build GRCh38
rs1005533271 Chr.16: 88428970-88428970 on Build GRCh38

View Full Product Details