Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52415266-52415537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF528
Entrez Gene ID
84436
Gene Name
zinc finger protein 528
Gene Aliases
-
UniGene
Hs.662043
Gene Chromosome Location
Chr.19: 52392771-52418408 on Build GRCh38
Transcripts
NM_032423.2 XM_005259323.1 XM_006723418.2 XM_011527392.1 XM_011527394.2 XM_017027360.1 XM_017027361.1 XM_017027362.1 XM_017027363.1 XM_017027364.1 XM_017027365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3404524 c.663C>G p.V221V Chr.19: 52415515-52415515 on Build GRCh38
3404523 c.604A>C p.T202P Chr.19: 52415456-52415456 on Build GRCh38
4900350 c.618C>T p.I206I Chr.19: 52415470-52415470 on Build GRCh38
4777434 c.431C>T p.S144L Chr.19: 52415283-52415283 on Build GRCh38
5936564 c.491G>A p.R164K Chr.19: 52415343-52415343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370434641 Chr.19: 52415477-52415477 on Build GRCh38
rs199532990 Chr.19: 52415505-52415505 on Build GRCh38
rs747344805 Chr.19: 52415363-52415363 on Build GRCh38
rs756399528 Chr.19: 52415492-52415492 on Build GRCh38
rs140617345 Chr.19: 52415317-52415317 on Build GRCh38
rs768765804 Chr.19: 52415368-52415368 on Build GRCh38
rs751608568 Chr.19: 52415479-52415479 on Build GRCh38
rs748261663 Chr.19: 52415381-52415381 on Build GRCh38
rs755578704 Chr.19: 52415314-52415314 on Build GRCh38
rs759305440 Chr.19: 52415425-52415425 on Build GRCh38
rs1021141094 Chr.19: 52415445-52415445 on Build GRCh38
rs137980249 Chr.19: 52415301-52415301 on Build GRCh38
rs903436457 Chr.19: 52415394-52415394 on Build GRCh38
rs750972115 Chr.19: 52415329-52415329 on Build GRCh38
rs761076852 Chr.19: 52415350-52415350 on Build GRCh38
rs562993017 Chr.19: 52415369-52415369 on Build GRCh38
rs760856744 Chr.19: 52415293-52415293 on Build GRCh38
rs370914123 Chr.19: 52415291-52415291 on Build GRCh38
rs760291956 Chr.19: 52415438-52415438 on Build GRCh38
rs766126846 Chr.19: 52415290-52415290 on Build GRCh38
rs377378031 Chr.19: 52415441-52415441 on Build GRCh38
rs959875855 Chr.19: 52415504-52415504 on Build GRCh38
rs758943294 Chr.19: 52415342-52415342 on Build GRCh38
rs767185559 Chr.19: 52415434-52415434 on Build GRCh38
rs774030096 Chr.19: 52415417-52415417 on Build GRCh38
rs779416983 Chr.19: 52415326-52415326 on Build GRCh38
rs893934248 Chr.19: 52415415-52415415 on Build GRCh38
rs778057102 Chr.19: 52415493-52415493 on Build GRCh38
rs753029876 Chr.19: 52415292-52415292 on Build GRCh38
rs766697481 Chr.19: 52415479-52415479 on Build GRCh38
rs781525871 Chr.19: 52415370-52415370 on Build GRCh38
rs750876390 Chr.19: 52415331-52415334 on Build GRCh38
rs900400865 Chr.19: 52415393-52415393 on Build GRCh38
rs758846335 Chr.19: 52415454-52415454 on Build GRCh38
rs753379000 Chr.19: 52415450-52415450 on Build GRCh38
rs761729829 Chr.19: 52415298-52415298 on Build GRCh38
rs773393696 Chr.19: 52415399-52415399 on Build GRCh38
rs571724070 Chr.19: 52415472-52415472 on Build GRCh38
rs547069115 Chr.19: 52415437-52415437 on Build GRCh38
rs766843246 Chr.19: 52415464-52415464 on Build GRCh38
rs768040541 Chr.19: 52415324-52415325 on Build GRCh38
rs200341438 Chr.19: 52415406-52415406 on Build GRCh38
rs762864246 Chr.19: 52415400-52415400 on Build GRCh38
rs1043489044 Chr.19: 52415390-52415390 on Build GRCh38
rs755321655 Chr.19: 52415476-52415476 on Build GRCh38
rs780344984 Chr.19: 52415358-52415358 on Build GRCh38

View Full Product Details