Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43543407-43543636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRCC1
Entrez Gene ID
7515
Gene Name
X-ray repair cross complementing 1
Gene Aliases
RCC
UniGene
Hs.98493
Gene Chromosome Location
Chr.19: 43543312-43575578 on Build GRCh38
Transcripts
NM_006297.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3823312 c.1794G>C p.L598L Chr.19: 43543500-43543500 on Build GRCh38
3823311 c.1870C>T p.H624Y Chr.19: 43543424-43543424 on Build GRCh38
7416573 c.1790C>A p.A597D Chr.19: 43543504-43543504 on Build GRCh38
1394287 c.1819G>A p.V607I Chr.19: 43543475-43543475 on Build GRCh38
7403208 c.1857G>C p.Q619H Chr.19: 43543437-43543437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749106716 Chr.19: 43543530-43543530 on Build GRCh38
rs751391164 Chr.19: 43543531-43543531 on Build GRCh38
rs751612285 Chr.19: 43543581-43543581 on Build GRCh38
rs750461941 Chr.19: 43543481-43543481 on Build GRCh38
rs911675465 Chr.19: 43543481-43543495 on Build GRCh38
rs3213400 Chr.19: 43543545-43543545 on Build GRCh38
rs755191942 Chr.19: 43543583-43543583 on Build GRCh38
rs745349091 Chr.19: 43543535-43543535 on Build GRCh38
rs777015591 Chr.19: 43543528-43543530 on Build GRCh38
rs199876205 Chr.19: 43543472-43543472 on Build GRCh38
rs922437146 Chr.19: 43543528-43543529 on Build GRCh38
rs765735154 Chr.19: 43543476-43543476 on Build GRCh38
rs377359870 Chr.19: 43543578-43543578 on Build GRCh38
rs189634883 Chr.19: 43543544-43543544 on Build GRCh38
rs771611144 Chr.19: 43543439-43543439 on Build GRCh38
rs777661657 Chr.19: 43543515-43543515 on Build GRCh38
rs3213399 Chr.19: 43543555-43543555 on Build GRCh38
rs760786423 Chr.19: 43543466-43543466 on Build GRCh38
rs759153919 Chr.19: 43543430-43543430 on Build GRCh38
rs368112519 Chr.19: 43543553-43543553 on Build GRCh38
rs773401514 Chr.19: 43543449-43543450 on Build GRCh38
rs762230089 Chr.19: 43543556-43543556 on Build GRCh38
rs753674953 Chr.19: 43543606-43543606 on Build GRCh38
rs921238159 Chr.19: 43543478-43543478 on Build GRCh38
rs767855212 Chr.19: 43543603-43543603 on Build GRCh38
rs755281928 Chr.19: 43543504-43543504 on Build GRCh38
rs754226766 Chr.19: 43543475-43543475 on Build GRCh38
rs371465894 Chr.19: 43543564-43543564 on Build GRCh38
rs1022665812 Chr.19: 43543440-43543440 on Build GRCh38
rs1048612955 Chr.19: 43543528-43543528 on Build GRCh38
rs766408364 Chr.19: 43543491-43543492 on Build GRCh38
rs757167693 Chr.19: 43543612-43543612 on Build GRCh38
rs780113067 Chr.19: 43543491-43543491 on Build GRCh38
rs3213398 Chr.19: 43543579-43543579 on Build GRCh38
rs201761108 Chr.19: 43543508-43543508 on Build GRCh38
rs751846839 Chr.19: 43543500-43543500 on Build GRCh38
rs771652590 Chr.19: 43543543-43543543 on Build GRCh38
rs184215643 Chr.19: 43543532-43543532 on Build GRCh38
rs145342527 Chr.19: 43543471-43543471 on Build GRCh38
rs548397667 Chr.19: 43543486-43543486 on Build GRCh38
rs369086567 Chr.19: 43543559-43543559 on Build GRCh38
rs144868354 Chr.19: 43543571-43543571 on Build GRCh38

View Full Product Details