Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89935503-89935762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBB3
Entrez Gene ID
10381
Gene Name
tubulin beta 3 class III
Gene Aliases
CDCBM, CDCBM1, CFEOM3, CFEOM3A, FEOM3, TUBB4, beta-4
UniGene
Hs.511743
Gene Chromosome Location
Chr.16: 89922009-89936097 on Build GRCh38
Transcripts
NM_001197181.1 NM_006086.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
290548 c.1293G>T p.E431D Chr.16: 89935744-89935744 on Build GRCh38
5094354 c.1281C>T p.D427D Chr.16: 89935732-89935732 on Build GRCh38
975023 c.1112G>A p.S371N Chr.16: 89935563-89935563 on Build GRCh38
5885699 c.1107G>C p.G369G Chr.16: 89935558-89935558 on Build GRCh38
5074684 c.1282G>A p.A428T Chr.16: 89935733-89935733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905888987 Chr.16: 89935647-89935647 on Build GRCh38
rs770275598 Chr.16: 89935708-89935708 on Build GRCh38
rs141064323 Chr.16: 89935666-89935666 on Build GRCh38
rs765882505 Chr.16: 89935603-89935603 on Build GRCh38
rs773849361 Chr.16: 89935609-89935609 on Build GRCh38
rs758588994 Chr.16: 89935678-89935678 on Build GRCh38
rs748724315 Chr.16: 89935705-89935705 on Build GRCh38
rs864622009 Chr.16: 89935739-89935739 on Build GRCh38
rs886042295 Chr.16: 89935620-89935620 on Build GRCh38
rs267607164 Chr.16: 89935700-89935700 on Build GRCh38
rs775768954 Chr.16: 89935569-89935569 on Build GRCh38
rs776916990 Chr.16: 89935702-89935702 on Build GRCh38
rs779199751 Chr.16: 89935663-89935663 on Build GRCh38
rs529172770 Chr.16: 89935527-89935527 on Build GRCh38
rs267607165 Chr.16: 89935679-89935679 on Build GRCh38
rs566062002 Chr.16: 89935567-89935567 on Build GRCh38
rs773898093 Chr.16: 89935710-89935710 on Build GRCh38
rs144689076 Chr.16: 89935651-89935651 on Build GRCh38
rs386794162 Chr.16: 89935711-89935712 on Build GRCh38
rs777734748 Chr.16: 89935528-89935528 on Build GRCh38
rs886039497 Chr.16: 89935623-89935623 on Build GRCh38
rs587784503 Chr.16: 89935621-89935621 on Build GRCh38
rs754560439 Chr.16: 89935522-89935522 on Build GRCh38
rs140825096 Chr.16: 89935531-89935531 on Build GRCh38
rs747151023 Chr.16: 89935696-89935696 on Build GRCh38
rs757440841 Chr.16: 89935555-89935555 on Build GRCh38
rs372849008 Chr.16: 89935732-89935732 on Build GRCh38
rs878853279 Chr.16: 89935613-89935613 on Build GRCh38
rs749198045 Chr.16: 89935529-89935529 on Build GRCh38
rs759051850 Chr.16: 89935711-89935711 on Build GRCh38
rs757192357 Chr.16: 89935657-89935657 on Build GRCh38
rs777149755 Chr.16: 89935590-89935590 on Build GRCh38
rs768863709 Chr.16: 89935582-89935582 on Build GRCh38
rs371418131 Chr.16: 89935648-89935648 on Build GRCh38
rs964422136 Chr.16: 89935742-89935742 on Build GRCh38
rs539745698 Chr.16: 89935576-89935576 on Build GRCh38
rs200882568 Chr.16: 89935737-89935737 on Build GRCh38
rs11554953 Chr.16: 89935597-89935597 on Build GRCh38
rs369375632 Chr.16: 89935699-89935699 on Build GRCh38
rs762254443 Chr.16: 89935598-89935598 on Build GRCh38
rs11554951 Chr.16: 89935534-89935534 on Build GRCh38
rs780355761 Chr.16: 89935684-89935684 on Build GRCh38
rs864321717 Chr.16: 89935589-89935589 on Build GRCh38
rs761071086 Chr.16: 89935580-89935580 on Build GRCh38
rs200607346 Chr.16: 89935741-89935741 on Build GRCh38
rs149337680 Chr.16: 89935634-89935634 on Build GRCh38
rs142238093 Chr.16: 89935738-89935738 on Build GRCh38

View Full Product Details