Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 51275731-51275964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP18
Entrez Gene ID
51096
Gene Name
UTP18, small subunit processome component
Gene Aliases
CGI-48, WDR50
UniGene
Hs.709327
Gene Chromosome Location
Chr.17: 51260536-51297933 on Build GRCh38
Transcripts
NM_016001.2 XM_006721930.3 XM_011524870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6081453 c.741A>T p.E247D Chr.17: 51275895-51275895 on Build GRCh38
4443306 c.720C>A p.N240K Chr.17: 51275874-51275874 on Build GRCh38
4739990 c.757C>T p.R253W Chr.17: 51275911-51275911 on Build GRCh38
3712386 c.732G>A p.A244A Chr.17: 51275886-51275886 on Build GRCh38
4508059 c.764C>G p.S255* Chr.17: 51275918-51275918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755417129 Chr.17: 51275902-51275902 on Build GRCh38
rs201600029 Chr.17: 51275938-51275938 on Build GRCh38
rs752238123 Chr.17: 51275838-51275838 on Build GRCh38
rs755327193 Chr.17: 51275825-51275825 on Build GRCh38
rs770398908 Chr.17: 51275837-51275837 on Build GRCh38
rs751990181 Chr.17: 51275820-51275820 on Build GRCh38
rs761704482 Chr.17: 51275869-51275869 on Build GRCh38
rs533877749 Chr.17: 51275911-51275911 on Build GRCh38
rs781772564 Chr.17: 51275832-51275832 on Build GRCh38
rs116854095 Chr.17: 51275769-51275769 on Build GRCh38
rs773331816 Chr.17: 51275871-51275871 on Build GRCh38
rs766628854 Chr.17: 51275886-51275886 on Build GRCh38
rs760624310 Chr.17: 51275942-51275943 on Build GRCh38
rs892749380 Chr.17: 51275936-51275936 on Build GRCh38
rs771768895 Chr.17: 51275846-51275846 on Build GRCh38
rs547487912 Chr.17: 51275912-51275912 on Build GRCh38
rs189542925 Chr.17: 51275818-51275818 on Build GRCh38
rs745563374 Chr.17: 51275845-51275845 on Build GRCh38
rs970042914 Chr.17: 51275797-51275797 on Build GRCh38
rs751792481 Chr.17: 51275896-51275896 on Build GRCh38
rs775732765 Chr.17: 51275929-51275929 on Build GRCh38
rs1023482035 Chr.17: 51275789-51275789 on Build GRCh38
rs549281509 Chr.17: 51275756-51275756 on Build GRCh38
rs768388026 Chr.17: 51275862-51275862 on Build GRCh38
rs929945856 Chr.17: 51275795-51275795 on Build GRCh38
rs531960072 Chr.17: 51275775-51275775 on Build GRCh38
rs765303641 Chr.17: 51275870-51275870 on Build GRCh38
rs912623111 Chr.17: 51275824-51275824 on Build GRCh38
rs756629212 Chr.17: 51275937-51275937 on Build GRCh38
rs774938630 Chr.17: 51275855-51275855 on Build GRCh38
rs914756354 Chr.17: 51275854-51275854 on Build GRCh38
rs181154558 Chr.17: 51275866-51275866 on Build GRCh38
rs745904625 Chr.17: 51275868-51275870 on Build GRCh38
rs745388460 Chr.17: 51275940-51275940 on Build GRCh38
rs748618939 Chr.17: 51275834-51275834 on Build GRCh38
rs772149350 Chr.17: 51275927-51275927 on Build GRCh38
rs762888616 Chr.17: 51275885-51275885 on Build GRCh38
rs760490086 Chr.17: 51275859-51275859 on Build GRCh38

View Full Product Details