Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124972038-124972269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZRANB1
Entrez Gene ID
54764
Gene Name
zinc finger RANBP2-type containing 1
Gene Aliases
TRABID
UniGene
Hs.595158
Gene Chromosome Location
Chr.10: 124916896-124988059 on Build GRCh38
Transcripts
NM_017580.2 XM_005269925.4 XM_005269926.4 XM_006717907.2 XM_017016357.1 XM_017016358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6834148 c.1153G>A p.A385T Chr.10: 124972115-124972115 on Build GRCh38
7141802 c.1125G>T p.L375L Chr.10: 124972087-124972087 on Build GRCh38
4776478 c.1149G>T p.L383F Chr.10: 124972111-124972111 on Build GRCh38
1234795 c.1105G>A p.D369N Chr.10: 124972067-124972067 on Build GRCh38
5962472 c.1118A>G p.Y373C Chr.10: 124972080-124972080 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs574837065 Chr.10: 124972094-124972094 on Build GRCh38
rs759895110 Chr.10: 124972144-124972144 on Build GRCh38
rs760177512 Chr.10: 124972133-124972133 on Build GRCh38
rs757424753 Chr.10: 124972164-124972164 on Build GRCh38
rs998166406 Chr.10: 124972215-124972215 on Build GRCh38
rs780125749 Chr.10: 124972102-124972102 on Build GRCh38
rs761540211 Chr.10: 124972164-124972165 on Build GRCh38
rs777898452 Chr.10: 124972158-124972159 on Build GRCh38
rs781352358 Chr.10: 124972167-124972167 on Build GRCh38
rs377041273 Chr.10: 124972066-124972066 on Build GRCh38
rs749120488 Chr.10: 124972108-124972108 on Build GRCh38
rs201266237 Chr.10: 124972166-124972167 on Build GRCh38
rs187107162 Chr.10: 124972099-124972099 on Build GRCh38
rs867554010 Chr.10: 124972119-124972119 on Build GRCh38
rs758430289 Chr.10: 124972126-124972126 on Build GRCh38
rs771686390 Chr.10: 124972128-124972128 on Build GRCh38
rs763625220 Chr.10: 124972162-124972163 on Build GRCh38
rs138810144 Chr.10: 124972137-124972137 on Build GRCh38
rs764407410 Chr.10: 124972163-124972163 on Build GRCh38
rs748201700 Chr.10: 124972146-124972147 on Build GRCh38
rs747063654 Chr.10: 124972085-124972085 on Build GRCh38
rs765666693 Chr.10: 124972158-124972158 on Build GRCh38
rs758696539 Chr.10: 124972160-124972160 on Build GRCh38
rs758485617 Chr.10: 124972147-124972148 on Build GRCh38
rs752970300 Chr.10: 124972078-124972078 on Build GRCh38
rs772744147 Chr.10: 124972130-124972130 on Build GRCh38
rs758509775 Chr.10: 124972079-124972079 on Build GRCh38
rs752957305 Chr.10: 124972159-124972159 on Build GRCh38
rs778730267 Chr.10: 124972124-124972125 on Build GRCh38
rs567307412 Chr.10: 124972136-124972136 on Build GRCh38
rs769649558 Chr.10: 124972160-124972161 on Build GRCh38
rs768476303 Chr.10: 124972106-124972106 on Build GRCh38
rs768501387 Chr.10: 124972117-124972117 on Build GRCh38
rs3802700 Chr.10: 124972212-124972212 on Build GRCh38
rs747856516 Chr.10: 124972127-124972127 on Build GRCh38
rs778022551 Chr.10: 124972080-124972080 on Build GRCh38
rs78649529 Chr.10: 124972221-124972221 on Build GRCh38
rs762416894 Chr.10: 124972161-124972162 on Build GRCh38
rs745637101 Chr.10: 124972125-124972126 on Build GRCh38

View Full Product Details