Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37725951-37726182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIOBP
Entrez Gene ID
11078
Gene Name
TRIO and F-actin binding protein
Gene Aliases
DFNB28, HRIHFB2122, TAP68, TARA, dJ37E16.4
UniGene
Hs.533030
Gene Chromosome Location
Chr.22: 37696988-37776556 on Build GRCh38
Transcripts
NM_001039141.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1034046 c.3549G>T p.E1183D Chr.22: 37726105-37726105 on Build GRCh38
3964269 c.3422G>A p.R1141K Chr.22: 37725978-37725978 on Build GRCh38
6808779 c.3415C>T p.L1139F Chr.22: 37725971-37725971 on Build GRCh38
6047030 c.3572C>T p.P1191L Chr.22: 37726128-37726128 on Build GRCh38
3150748 c.3506G>A p.R1169Q Chr.22: 37726062-37726062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368198510 Chr.22: 37726007-37726007 on Build GRCh38
rs769650134 Chr.22: 37725982-37725982 on Build GRCh38
rs5756795 Chr.22: 37726115-37726115 on Build GRCh38
rs898100103 Chr.22: 37725994-37725994 on Build GRCh38
rs1017691527 Chr.22: 37726043-37726043 on Build GRCh38
rs368562914 Chr.22: 37726142-37726142 on Build GRCh38
rs781152926 Chr.22: 37725973-37725973 on Build GRCh38
rs746393533 Chr.22: 37726100-37726100 on Build GRCh38
rs866151427 Chr.22: 37726085-37726085 on Build GRCh38
rs774652028 Chr.22: 37726128-37726128 on Build GRCh38
rs1032335712 Chr.22: 37726046-37726046 on Build GRCh38
rs749084232 Chr.22: 37726064-37726064 on Build GRCh38
rs763273357 Chr.22: 37726125-37726125 on Build GRCh38
rs773132022 Chr.22: 37725984-37725984 on Build GRCh38
rs764276281 Chr.22: 37726041-37726041 on Build GRCh38
rs756181068 Chr.22: 37726153-37726153 on Build GRCh38
rs780058028 Chr.22: 37726062-37726062 on Build GRCh38
rs34095863 Chr.22: 37726147-37726148 on Build GRCh38
rs372266271 Chr.22: 37726112-37726112 on Build GRCh38
rs200646548 Chr.22: 37726021-37726021 on Build GRCh38
rs374418895 Chr.22: 37725995-37725995 on Build GRCh38
rs866435996 Chr.22: 37726117-37726117 on Build GRCh38
rs765602011 Chr.22: 37726024-37726024 on Build GRCh38
rs866948221 Chr.22: 37726155-37726155 on Build GRCh38
rs751630433 Chr.22: 37726049-37726049 on Build GRCh38
rs771696726 Chr.22: 37726012-37726013 on Build GRCh38
rs767442889 Chr.22: 37726055-37726055 on Build GRCh38
rs747718808 Chr.22: 37726084-37726084 on Build GRCh38
rs771635907 Chr.22: 37726092-37726092 on Build GRCh38
rs764400987 Chr.22: 37726127-37726127 on Build GRCh38
rs370721352 Chr.22: 37725985-37725985 on Build GRCh38
rs1000398077 Chr.22: 37726044-37726044 on Build GRCh38
rs754855665 Chr.22: 37726162-37726162 on Build GRCh38
rs538294501 Chr.22: 37726156-37726156 on Build GRCh38
rs867123030 Chr.22: 37726150-37726150 on Build GRCh38
rs771386143 Chr.22: 37726012-37726012 on Build GRCh38
rs751210935 Chr.22: 37726154-37726154 on Build GRCh38
rs775766196 Chr.22: 37726030-37726030 on Build GRCh38
rs778600630 Chr.22: 37726077-37726077 on Build GRCh38
rs956274112 Chr.22: 37726068-37726068 on Build GRCh38
rs750461919 Chr.22: 37726144-37726144 on Build GRCh38
rs757388800 Chr.22: 37726051-37726051 on Build GRCh38
rs201840128 Chr.22: 37726074-37726074 on Build GRCh38
rs756046866 Chr.22: 37726061-37726061 on Build GRCh38
rs867108679 Chr.22: 37726083-37726084 on Build GRCh38
rs199656357 Chr.22: 37726163-37726163 on Build GRCh38
rs763218276 Chr.22: 37726038-37726038 on Build GRCh38
rs747677122 Chr.22: 37726011-37726011 on Build GRCh38
rs748950216 Chr.22: 37725991-37725991 on Build GRCh38
rs867570366 Chr.22: 37726073-37726073 on Build GRCh38
rs761879051 Chr.22: 37726135-37726135 on Build GRCh38
rs772539880 Chr.22: 37726093-37726093 on Build GRCh38
rs745749363 Chr.22: 37725980-37725980 on Build GRCh38
rs759911435 Chr.22: 37726022-37726022 on Build GRCh38
rs750383590 Chr.22: 37726060-37726060 on Build GRCh38

View Full Product Details